Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01251786_s1
          See other KCNK15 GE Assays ›
          Gene Symbol:
          Kcnk15
          Gene Name:
          potassium channel, subfamily K, member 15
          Gene Aliases:
          KCNK11, KCNK14, KT3.3, TASK5
          Chromosome Location:
          Chr.2: 163853750 - 163858874 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01251786_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnk15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001030292.1 NP_001025463.1 2 466 103
          GenBank mRNA BC147255.1 - 2 565 9056306 103
          BC147256.1 - 2 565 9056288 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002497.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnk15

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          stabilization of membrane potential
          potassium ion transmembrane transport
          potassium channel activity
          potassium ion leak channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21814792 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21814761 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21814797 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21814796 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21814760 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21814795 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21814762 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156006 NM_001030292.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156006 BC147255.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156006 BC147256.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568052 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 279 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 190519 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156006 NM_001030292.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156006 BC147255.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156006 BC147256.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21814759 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04770624 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01701449 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01701455 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21814762 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21814760 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21814761 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21814795 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21814797 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21814792 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21814796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21814775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01172446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01148982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01974270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02116036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01701455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02280923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01974284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01172448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01974257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01869383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01172449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01974254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01148981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02103948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01148987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01148990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21814795 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10478423 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478422 - -
          Affymetrix HT MG-430 PM Array Plate 1447972_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10478421 - -
          Mouse Genome 430 2.0 Array 1447972_at - -
          Mouse Genome 430 2.0 Array 1447972_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200020387.mm.1 TC0200002497.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020386.mm.1 TC0200002497.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020386.mm.1 NM_001030292.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020386.mm.1 NM_001030292.1 Refseq
          Mouse Expression Set 430 1447972_PM_at - -
          Mouse Expression Set 430 1447972_at - -
          Affymetrix HT Mouse Genome 430B Array 1447972_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447972_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 582106 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230948 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3045874 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3121407 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3121407 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3045874 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0