Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01252544_m1
          See other SPHK1 GE Assays ›
          Gene Symbol:
          Sphk1
          Gene Name:
          sphingosine kinase 1
          Gene Aliases:
          1110006G24Rik, Sk1, Spk1
          Chromosome Location:
          Chr.11: 116529817 - 116536675 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01252544_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sphk1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001172472.1 NP_001165943.1 2 - 3 449 97
          NM_001172473.1 NP_001165944.1 2 - 3 335 97
          NM_025367.6 NP_079643.2 2 - 3 332 97
          GenBank mRNA AK155332.1 - 2 - 3 434 97
          AK160900.1 - 2 - 3 325 97
          BC037710.1 - 2 - 3 247 4221357 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001935.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK155332.1, AK160900.1, BC037710.1, NM_001172472.1, NM_001172473.1, NM_025367.6 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sphk1

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          positive regulation of protein phosphorylation
          positive regulation of neurotransmitter secretion
          sphingosine-1-phosphate receptor signaling pathway
          sphingosine metabolic process
          inflammatory response
          brain development
          positive regulation of cell population proliferation
          positive regulation of peptidyl-threonine phosphorylation
          regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of neuron projection development
          phosphorylation
          cyclooxygenase pathway
          calcium-mediated signaling
          positive regulation of cell growth
          positive regulation of cell migration
          positive regulation of protein ubiquitination
          regulation of interleukin-1 beta production
          negative regulation of apoptotic process
          positive regulation of angiogenesis
          positive regulation of mitotic cell cycle
          positive regulation of smooth muscle contraction
          sphingosine biosynthetic process
          lipid phosphorylation
          positive regulation of fibroblast proliferation
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of NIK/NF-kappaB signaling
          nucleotide binding
          magnesium ion binding
          DNA binding
          calmodulin binding
          ATP binding
          sphinganine kinase activity
          kinase activity
          transferase activity
          D-erythro-sphingosine kinase activity
          sphingosine-1-phosphate receptor activity
          protein phosphatase 2A binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100018369.mm.1 TC1100001935.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100018369.mm.1 NM_025367.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018369.mm.1 NM_025367.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018364.mm.1 TC1100001935.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100018364.mm.1 NM_001172472.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018364.mm.1 NM_001172472.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 339191 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-ffb8c786-92mpb:80/100.66.129.117:80.
          git-commit: 1a801765356e65be0b6ed8595ed79b52bb698fed
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0