Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01253238_g1
          See other ITGB1 GE Assays ›
          Gene Symbol:
          Itgb1
          Gene Name:
          integrin beta 1 (fibronectin receptor beta)
          Gene Aliases:
          4633401G24Rik, AA409975, AA960159, CD29, ENSMUSG00000051907, Fnrb, Gm9863, gpIIa
          Chromosome Location:
          Chr.8: 128685284 - 128733579 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01253238_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Itgb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010578.2 NP_034708.1 8 - 9 1344 124
          GenBank mRNA AK076526.1 - 4 - 5 641 124
          AK088016.1 - 8 - 9 1245 124
          AK088729.1 - 8 - 9 1091 124
          AK135521.1 - 7 - 8 1020 124
          AK141253.1 - 3 - 4 271 124
          AK159689.1 - 8 - 9 1186 124
          AK167682.1 - 8 - 9 1242 124
          BC050906.1 - 8 - 9 1217 5721348 124
          X15202.1 - 7 - 8 1037 124
          Y00769.1 - 8 - 9 1132 124

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Itgb1

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          in utero embryonic development
          cell fate specification
          tissue homeostasis
          cell migration involved in sprouting angiogenesis
          cellular calcium ion homeostasis
          cell adhesion
          leukocyte cell-cell adhesion
          cell-matrix adhesion
          calcium-independent cell-matrix adhesion
          transforming growth factor beta receptor signaling pathway
          integrin-mediated signaling pathway
          regulation of G protein-coupled receptor signaling pathway
          positive regulation of cell population proliferation
          negative regulation of cell population proliferation
          germ cell migration
          visual learning
          regulation of collagen catabolic process
          positive regulation of cell-substrate adhesion
          positive regulation of neuron projection development
          cell migration
          formation of radial glial scaffolds
          positive regulation of cell migration
          neuron projection development
          negative regulation of cell projection organization
          cell-substrate adhesion
          protein transport within lipid bilayer
          heterotypic cell-cell adhesion
          negative regulation of Rho protein signal transduction
          positive regulation of apoptotic process
          stress fiber assembly
          positive regulation of MAPK cascade
          sarcomere organization
          negative regulation of cell differentiation
          negative regulation of neuron differentiation
          positive regulation of neuron differentiation
          positive regulation of endocytosis
          axon extension
          cardiac muscle tissue development
          dendrite morphogenesis
          positive regulation of peptidyl-tyrosine phosphorylation
          leukocyte tethering or rolling
          regulation of cell cycle
          cardiac muscle cell differentiation
          bicellular tight junction assembly
          positive regulation of protein localization to plasma membrane
          negative regulation of anoikis
          fibronectin binding
          protease binding
          actin binding
          signaling receptor binding
          integrin binding
          protein binding
          collagen binding
          kinase binding
          protein kinase binding
          protein domain specific binding
          signaling receptor activity
          peptide binding
          laminin binding
          protein-containing complex binding
          metal ion binding
          protein heterodimerization activity
          cell adhesion molecule binding
          alpha-actinin binding
          collagen binding involved in cell-matrix adhesion

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0