Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01254911_g1
          See other GNPDA1 GE Assays ›
          Gene Symbol:
          Gnpda1 Gm8615
          Gene Name:
          glucosamine-6-phosphate deaminase 1
          glucosamine-6-phosphate deaminase 1 pseudogene
          Gene Aliases:
          GNPDA, Gnp1, Gnpi, oscillin
          EG667410
          Chromosome Location:
          Chr.18: 38327537 - 38338993 on Build GRCm38
          Chromosome Location:
          Chr.5: 149154128 - 149156192 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm01254911_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gnpda1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011937.2 NP_036067.2 4 - 5 502 115
          GenBank mRNA AF088903.1 - 4 - 5 460 115
          AF160355.1 - 4 - 5 448 115
          AK077625.1 - 1 - 2 167 115
          AK167088.1 - 1 - 2 176 115
          AK172879.1 - 3 - 4 365 115
          BC014800.1 - 4 - 5 433 4220980 115
          Gene Symbol: Gm8615
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028061.1 1 285 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003651.mm.1, TC1800001230.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gnpda1
          Click here to find siRNAs for Gene Gm8615

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          fructose 6-phosphate metabolic process
          glucosamine metabolic process
          glucosamine catabolic process
          N-acetylglucosamine metabolic process
          generation of precursor metabolites and energy
          acrosome reaction
          fructose biosynthetic process
          glucosamine-6-phosphate deaminase activity
          hydrolase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800009158.mm.1 TC1800001230.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009158.mm.1 NM_011937.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009158.mm.1 NM_011937.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009157.mm.1 TC1800001230.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009157.mm.1 NM_011937.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009157.mm.1 NM_011937.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009154.mm.1 TC1800001230.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009154.mm.1 NM_011937.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009154.mm.1 NM_011937.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033551.mm.1 TC0500003651.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033551.mm.1 NR_028061.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033552.mm.1 TC0500003651.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033552.mm.1 NR_028061.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009155.mm.1 TC1800001230.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009155.mm.1 NM_011937.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009155.mm.1 NM_011937.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0