Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198570.1 | NP_001185499.1 | 5 - 6 | 767 | 93 | |
NM_001198571.1 | NP_001185500.1 | 5 - 6 | 767 | 93 | ||
NM_198127.2 | NP_937760.1 | 4 - 5 | 749 | 93 | ||
GenBank mRNA | BC056345.1 | - | 4 - 5 | 570 | 5705048 | 93 |
BC079545.1 | - | 5 - 6 | 652 | 6851145 | 93 | |
BC079646.1 | - | 5 - 6 | 691 | 6850720 | 93 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |