Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100019431.mm.1 |
TC0100002385.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019431.mm.1 |
NM_172513.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019431.mm.1 |
NM_172513.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019433.mm.1 |
TC0100002385.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019433.mm.1 |
NM_172513.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019433.mm.1 |
NM_172513.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019430.mm.1 |
TC0100002385.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019430.mm.1 |
NM_172513.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019430.mm.1 |
NM_172513.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019432.mm.1 |
TC0100002385.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019432.mm.1 |
NM_172513.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019432.mm.1 |
NM_172513.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019429.mm.1 |
TC0100002385.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019429.mm.1 |
NM_172513.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019429.mm.1 |
NM_172513.3 |
Refseq |