Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01257282_m1
          See other STAT1 GE Assays ›
          Gene Symbol:
          Stat1
          Gene Name:
          signal transducer and activator of transcription 1
          Gene Aliases:
          2010005J02Rik, AA408197
          Chromosome Location:
          Chr.1: 52119438 - 52161865 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01257282_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stat1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001205313.1 NP_001192242.1 15 - 16 1560 81
          NM_001205314.1 NP_001192243.1 15 - 16 1596 81
          NM_009283.4 NP_033309.3 15 - 16 1611 81
          GenBank mRNA AK018544.1 - 15 - 16 1562 81
          AK039458.1 - 15 - 16 1612 81
          AK046517.1 - 15 - 16 1560 81
          AK082706.1 - 15 - 16 1593 81
          AK084855.1 - 15 - 16 1545 81
          AK157263.1 - 15 - 16 1460 81
          AK159887.1 - 15 - 16 1595 81
          BC004808.1 - 15 - 16 1547 3587831 81
          U06924.1 - 12 - 13 1265 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000375.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stat1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endothelial cell proliferation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          signal transduction
          receptor signaling pathway via JAK-STAT
          response to nutrient
          blood circulation
          positive regulation of cell population proliferation
          response to xenobiotic stimulus
          response to mechanical stimulus
          response to bacterium
          negative regulation of angiogenesis
          cytokine-mediated signaling pathway
          lipopolysaccharide-mediated signaling pathway
          response to lipopolysaccharide
          cellular response to insulin stimulus
          tumor necrosis factor-mediated signaling pathway
          response to cytokine
          negative regulation of macrophage fusion
          cellular response to interferon-beta
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          response to exogenous dsRNA
          response to peptide hormone
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          negative regulation by virus of viral protein levels in host cell
          positive regulation of smooth muscle cell proliferation
          response to cAMP
          interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          renal tubule development
          cellular response to lipopolysaccharide
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          DNA binding
          double-stranded DNA binding
          DNA-binding transcription factor activity
          tumor necrosis factor receptor binding
          protein binding
          enzyme binding
          CCR5 chemokine receptor binding
          identical protein binding
          protein homodimerization activity
          sequence-specific DNA binding
          protein phosphatase 2A binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100002908.mm.1 TC0100000375.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100002908.mm.1 NM_001205313.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002908.mm.1 NM_001205313.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002908.mm.1 NM_001205314.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002908.mm.1 NM_001205314.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002908.mm.1 NM_009283.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002908.mm.1 NM_009283.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002909.mm.1 TC0100000375.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100002909.mm.1 NM_001205313.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002909.mm.1 NM_001205313.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002909.mm.1 NM_001205314.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002909.mm.1 NM_001205314.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002909.mm.1 NM_009283.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002909.mm.1 NM_009283.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002911.mm.1 TC0100000375.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100002911.mm.1 NM_001205313.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002911.mm.1 NM_001205313.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002911.mm.1 NM_001205314.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002911.mm.1 NM_001205314.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002911.mm.1 NM_009283.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002911.mm.1 NM_009283.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0