Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01259432_s1
          See other 1700106J12RIK GE Assays ›
          Gene Symbol:
          1700106J12Rik
          Gene Name:
          RIKEN cDNA 1700106J12 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01259432_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700106J12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK007128.1 - 1 741 75
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB015462.1 - 1 154 Chr. 19 - 56066714 - 56066926 [+] on Build GRCm38 75
          GenBankmRNA AI449781.1 - 1 Chr. 19 - 56066484 - 56066928 [-] on Build GRCm38 75

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transcription factor 7 like 2,
          T cell specific,
          HMG box
          Tcf7l2 TCF4B,
          TCF4E,
          Tcf-4,
          Tcf4,
          mTcf-4B,
          mTcf-4E
          -132.8kb Anti-sense Chr. 19 - 55741810 - 55933661 on NCBI Build GRCm38
          Downstream predicted gene,
          31912
          Gm31912 38.8kb Anti-sense Chr. 19 - 56105689 - 56109342 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14495832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14322118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14489197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14157661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14495814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14385336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14341094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14174943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14385842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14341099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14318971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14141557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14279240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14157665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14495829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14385847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32198377 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1457006_PM_at - -
          Mouse Genome 430 2.0 Array 1457006_at - -
          Mouse Genome 430 2.0 Array 1457006_PM_at - -
          Mouse Expression Set 430 1457006_PM_at - -
          Mouse Expression Set 430 1457006_at - -
          Affymetrix HT Mouse Genome 430B Array 1457006_at - -
          Affymetrix HT Mouse Genome 430B Array 1457006_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 747165 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 107040397 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-ffb8c786-kqswr:80/100.66.130.62:80.
          git-commit: 1a801765356e65be0b6ed8595ed79b52bb698fed
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0