Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01259439_g1
          See other 1810007D17RIK GE Assays ›
          Gene Symbol:
          1810007D17Rik
          Gene Name:
          RIKEN cDNA 1810007D17 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 58606329 - 58629753 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01259439_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1810007D17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038136.1 2 - 3 327 78
          GenBank mRNA AK007359.1 - 2 - 3 331 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream coiled-coil domain containing 172 Ccdc172 1700011F14Rik -75.8kb Sense Chr. 19 - 58511999 - 58553104 on NCBI Build GRCm38
          Downstream pancreatic lipase Pnlip 1810007A24Rik,
          PL,
          PTL
          40.6kb Sense Chr. 19 - 58670365 - 58681788 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000837.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2309020 AK007359.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 38541 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 506602 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 872315 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 130924 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755843 AF200495.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755843 AF230378.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755843 AJ250429.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755843 AK008977.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755843 AK009741.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755843 AK010020.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755843 AK014576.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755843 BC132600.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755843 BC132602.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755843 NM_019451.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 17825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169856 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 9260 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 138604 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155847 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2309020 AK007359.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155847 - -
          Whole Mouse Genome G4122A A_52_P1106665 AK007359.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139931 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32214414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32214471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14317025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14217562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32214409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32214412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32214410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32214469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32214407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32214404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14210305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32214419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14167619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32214417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32214418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32214415 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1456198_PM_at - -
          Mouse Genome 430 2.0 Array 1456198_at - -
          Mouse Genome 430 2.0 Array 1456198_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900007275.mm.1 TC1900000837.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900007275.mm.1 NR_038136.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007275.mm.1 NR_038136.1 Refseq
          Mouse Expression Set 430 1456198_at - -
          Mouse Expression Set 430 1456198_PM_at - -
          Murine Genome U74v2 Set 165923_r_at - -
          Murine Genome U74v2 Set 165923_PM_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1456198_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456198_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 435501 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101850368 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline