Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01259578_m1
          See other SOX5OS3 GE Assays ›
          Gene Symbol:
          Sox5os3
          Gene Name:
          SRY (sex determining region Y)-box 5, opposite strand 3
          Gene Aliases:
          4933425H06Rik
          Chromosome Location:
          Chr.6: 144672868 - 144693832 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01259578_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sox5os3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040519.1 2 - 3 328 66
          GenBank mRNA AK006194.1 - 2 - 3 329 66
          AK016914.1 - 2 - 3 314 66
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CJ132009.1 - 2 - 3 437 Chr. 6 - 144679702 - 144693584 [+] on Build GRCm38 66
          GenBankmRNA BY716593.1 - 2 - 3 314 Chr. 6 - 144672882 - 144693835 [+] on Build GRCm38 66
          GenBankmRNA BY706291.1 - 2 - 3 329 Chr. 6 - 144672868 - 144693573 [+] on Build GRCm38 66
          GenBankmRNA BU961281.1 - 2 - 3 303 Chr. 6 - 144672896 - 144693569 [+] on Build GRCm38 66
          GenBankmRNA CN834653.1 - 330 Chr. - - [-] on Build 66

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream SRY (sex determining region Y)-box 5 Sox5 A730017D01Rik,
          AI528773
          Anti-sense Chr. 6 - 143828425 - 144781989 on NCBI Build GRCm38
          Downstream branched chain aminotransferase 1,
          cytosolic
          Bcat1 BCATc,
          Eca39
          300.3kb Anti-sense Chr. 6 - 144993835 - 145076157 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001703.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P574158 AK006194.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P574158 AK016914.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P574158 AK006194.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P574158 AK016914.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P574158 AK006194.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P574158 AK016914.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P574158 AK006194.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P574158 AK016914.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P574158 AK006194.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P574158 AK016914.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P574158 AK006194.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P574158 AK016914.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05319602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05450111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05061414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05749488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05369014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05450072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05061392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05391225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05319605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24918630 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600013541.mm.1 TC0600001703.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600013541.mm.1 NR_040519.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600013541.mm.1 NR_040519.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600013542.mm.1 TC0600001703.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600013542.mm.1 NR_040519.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600013542.mm.1 NR_040519.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 550441 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0