Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01259612_m1
          See other 4930528G23RIK GE Assays ›
          Gene Symbol:
          4930528G23Rik
          Gene Name:
          RIKEN cDNA 4930528G23 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 148659978 - 148691917 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01259612_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930528G23Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015927.1 - 3 - 4 795 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 6313 Gm6313 EG622319 -45.7kb Sense Chr. 6 - 148606786 - 148614309 on NCBI Build GRCm38
          Downstream importin 8 Ipo8 6230418K12Rik,
          Abcc10,
          C130009K11Rik,
          MRP7,
          OM-1,
          Om1,
          Ranbp8
          103.8kb Sense Chr. 6 - 148770683 - 148831467 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2277895 AK015927.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966009 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 655659 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 571156 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 761706 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 182770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 46097 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2277895 AK015927.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06109985 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01474087 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130880 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06109985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24941422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05384427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24941419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24941407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05384420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24941414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24941424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24941409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24941411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24941416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24941420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24941412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24941426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24941418 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431950_PM_at - -
          Mouse Genome 430 2.0 Array 1431950_at - -
          Mouse Genome 430 2.0 Array 1431950_PM_at - -
          Mouse Expression Set 430 1431950_PM_at - -
          Mouse Expression Set 430 1431950_at - -
          Affymetrix HT Mouse Genome 430B Array 1431950_at - -
          Affymetrix HT Mouse Genome 430B Array 1431950_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 648520 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102680044 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0