Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400033769.mm.1 |
TC0400004062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400033769.mm.1 |
NM_009802.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033769.mm.1 |
NM_009802.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033771.mm.1 |
TC0400004062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400033771.mm.1 |
NM_009802.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033771.mm.1 |
NM_009802.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033770.mm.1 |
TC0400004062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400033770.mm.1 |
NM_009802.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033770.mm.1 |
NM_009802.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033773.mm.1 |
TC0400004062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400033773.mm.1 |
NM_009802.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033773.mm.1 |
NM_009802.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033775.mm.1 |
TC0400004062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400033775.mm.1 |
NM_009802.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033775.mm.1 |
NM_009802.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033772.mm.1 |
TC0400004062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400033772.mm.1 |
NM_009802.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033772.mm.1 |
NM_009802.2 |
Refseq |