Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285529.1 | NP_001272458.1 | 11 - 12 | 1790 | 85 | |
NM_001285530.1 | NP_001272459.1 | 9 - 10 | 1571 | 85 | ||
NM_011599.5 | NP_035729.3 | 11 - 12 | 1793 | 85 | ||
GenBank mRNA | AK048515.1 | - | 11 - 12 | 1780 | 85 | |
AK052961.1 | - | 9 - 10 | 1894 | 85 | ||
AK082499.1 | - | 11 - 12 | 1759 | 85 | ||
AK215048.1 | - | 1 - 2 | 52 | 85 | ||
AY155195.1 | - | 11 - 12 | 952 | 85 | ||
AY155197.1 | - | 9 - 10 | 729 | 85 | ||
AY155198.1 | - | 7 - 8 | 592 | 85 | ||
AY155199.1 | - | 11 - 12 | 952 | 85 | ||
AY155200.1 | - | 5 - 6 | 426 | 85 | ||
BC057573.1 | - | 11 - 12 | 1620 | 6401650 | 85 | |
U61362.1 | - | 11 - 12 | 959 | 85 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |