Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01260693_m1
          See other AMN1 GE Assays ›
          Gene Symbol:
          Amn1
          Gene Name:
          antagonist of mitotic exit network 1
          Gene Aliases:
          5830467E07Rik, A230083D09, C730024G19Rik
          Chromosome Location:
          Chr.6: 149157349 - 149188762 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01260693_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Amn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113424.1 NP_001106895.1 6 - 7 767 155
          GenBank mRNA AK038995.1 - 4 - 5 813 155
          AK053764.1 - 3 - 4 2702 155
          BC026924.1 - 4 - 5 1950 155

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003482.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2454896 AK053764.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2454896 BC026924.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2454896 AK050169.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2454896 AK038995.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2454896 NM_001113424.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2366273 AK018037.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573859 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670082 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 141049 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813983 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529296 AF226662.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529296 AK044944.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529296 BC109167.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529296 BC109168.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529296 NM_033652.5 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793307 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858568 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 693614 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930739 AF148216.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930739 AF459018.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930739 AK019687.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930739 BC004637.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930739 BC093528.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930739 NM_001113356.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930739 NM_023143.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930739 XM_001002804.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930739 XM_001480171.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 803062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 788026 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299390 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 786850 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529741 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 936601 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831684 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 270993 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 22556 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 20993 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51591 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 19670 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131510 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2454896 AK053764.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2454896 BC026924.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2454896 AK050169.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2454896 AK038995.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2454896 NM_001113424.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06111215 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06111196 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06111160 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06111231 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06111178 - -
          Whole Mouse Genome G4122A A_52_P588262 AK053764.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P588262 AK050169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P588262 AK038995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P588262 NM_001113424.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P464592 AK050537.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P464592 AK036242.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06111160 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06111196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24944012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24944022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24944015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24944002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24944018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24944019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24944003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24944004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24944021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24944006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24944011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24943964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05776633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05821707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24944015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24944004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05390025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24944011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05325061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05174585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24944003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05444210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05575166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05048699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05776634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05553268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24944000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05784949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05840618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05307587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24943956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05325063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24944006 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10549538 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549541 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549543 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549540 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549539 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549537 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549542 - -
          Affymetrix HT MG-430 PM Array Plate 1453358_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1429978_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441141_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1454766_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10549536 - -
          Mouse Genome 430 2.0 Array 1454766_at - -
          Mouse Genome 430 2.0 Array 1453358_PM_s_at - -
          Mouse Genome 430 2.0 Array 1453358_s_at - -
          Mouse Genome 430 2.0 Array 1441141_PM_at - -
          Mouse Genome 430 2.0 Array 1441141_at - -
          Mouse Genome 430 2.0 Array 1454766_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232566_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232566_at - -
          Affymetrix Mouse Clariom S Array PSR0600027543.mm.1 TC0600003482.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600027543.mm.1 NM_001113424.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600027543.mm.1 NM_001113424.1 Refseq
          Mouse Expression Set 430 1454766_at - -
          Mouse Expression Set 430 1453358_s_at - -
          Mouse Expression Set 430 1454766_PM_at - -
          Mouse Expression Set 430 1453358_PM_s_at - -
          Mouse Expression Set 430 1441141_PM_at - -
          Mouse Expression Set 430 1441141_at - -
          Affymetrix HT Mouse Genome 430B Array 1454766_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454766_at - -
          Affymetrix HT Mouse Genome 430B Array 1441141_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441141_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 841266 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1217441 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217441 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0