Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01260973_g1
          See other 1700072G22RIK GE Assays ›
          Gene Symbol:
          1700072G22Rik
          Gene Name:
          RIKEN cDNA 1700072G22 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01260973_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700072G22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK006937.1 - 1 - 2 164 70
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY706952.1 - 1 - 2 159 Chr. 1 - 51457932 - 51458931 [-] on Build GRCm38 70

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 9330175M20 gene 9330175M20Rik -124.2kb Sense Chr. 1 - 51218386 - 51333779 on NCBI Build GRCm38
          Downstream nucleic acid binding protein 1 Nabp1 4930434H03Rik,
          4930442A21Rik,
          4930488J04Rik,
          4933440J18Rik,
          5830411E10Rik,
          AI852561,
          Nbp1,
          Obfc2a,
          Ssb2
          6.4kb Sense Chr. 1 - 51465297 - 51478458 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 376152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00971372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01135345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00734522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00377383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00971368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01106230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00971371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01145337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01145331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01145335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00734525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00726712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20221480 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 797965 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100070040 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0