Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01261032_g1
          See other 5830416P10RIK GE Assays ›
          Gene Symbol:
          5830416P10Rik
          Gene Name:
          RIKEN cDNA 5830416P10 gene
          Gene Aliases:
          4833407H14Rik, 5430437C04Rik, ENSMUSG00000074788, Gm10758
          Chromosome Location:
          Chr.19: 53441212 - 53464796 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01261032_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5830416P10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028427.1 4 - 5 530 162
          GenBank mRNA AK158034.1 - 4 - 5 518 162

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001672.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027708 AK158034.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027708 NR_028427.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419246 AK158034.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419246 AK019949.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419246 AK042769.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419246 AK047177.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419246 NR_028427.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419246 AK158034.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419246 AK019949.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419246 AK042769.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419246 AK047177.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419246 NR_028427.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765220 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 363907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365318 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 105754 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 745137 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886371 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13268 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 708735 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 72841 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71542 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8701 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 6742 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140162 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163543 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027708 AK158034.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027708 NR_028427.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419246 AK158034.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419246 AK019949.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419246 AK042769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419246 AK047177.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419246 NR_028427.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419246 AK158034.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419246 AK019949.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419246 AK042769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419246 AK047177.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419246 NR_028427.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P195987 AK079678.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P195987 AK017935.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P14350594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14254379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14195733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14255710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14245428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14190425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14415602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14190427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14439052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14254375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14460964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14203320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14354215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14392449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14460960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32182631 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453244_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10468527 - -
          Mouse Genome 430 2.0 Array 1453244_at - -
          Mouse Genome 430 2.0 Array 1453244_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900014749.mm.1 TC1900001672.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900014749.mm.1 NR_028427.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014749.mm.1 NR_028427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014742.mm.1 TC1900001672.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900014742.mm.1 NR_028427.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014742.mm.1 NR_028427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014739.mm.1 TC1900001672.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900014739.mm.1 NR_028427.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014739.mm.1 NR_028427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014740.mm.1 TC1900001672.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900014740.mm.1 NR_028427.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014740.mm.1 NR_028427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014751.mm.1 TC1900001672.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900014751.mm.1 NR_028427.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014751.mm.1 NR_028427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014752.mm.1 TC1900001672.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900014752.mm.1 NR_028427.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014752.mm.1 NR_028427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014743.mm.1 TC1900001672.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900014743.mm.1 NR_028427.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014743.mm.1 NR_028427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014734.mm.1 TC1900001672.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900014734.mm.1 NR_028427.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014734.mm.1 NR_028427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014750.mm.1 TC1900001672.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900014750.mm.1 NR_028427.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014750.mm.1 NR_028427.1 Refseq
          Mouse Expression Set 430 1453244_at - -
          Mouse Expression Set 430 1453244_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453244_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453244_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 763801 - -
          ABI Mouse Genome Survey Microarray 773688 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0