Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam204a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029648.6 | NP_083924.2 | 3 - 4 | 108 | ||
GenBank mRNA | AK075914.1 | - | 3 - 4 | 108 | ||
AK162079.1 | - | 3 - 4 | 108 | |||
BC061459.1 | - | 6829614 | 108 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |