Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01261972_g1
          See other TIMM21 GE Assays ›
          Gene Symbol:
          Timm21
          Gene Name:
          tranlocase of inner mitochondrial membrane 21
          Gene Aliases:
          1700034H14Rik, 2700002I20Rik
          Chromosome Location:
          Chr.18: 84947294 - 84951738 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01261972_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Timm21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025969.4 NP_080245.1 5 - 6 868 125
          GenBank mRNA AK006594.1 - 5 - 6 373 125
          AK012203.1 - 5 - 6 868 125
          AK013281.1 - 5 - 6 781 125
          AK017477.1 - 5 - 6 835 125
          AK029037.1 - 3 - 4 2045 125
          AK032468.1 - 3 - 4 2049 125
          AK168105.1 - 5 - 6 779 125
          BC034297.1 - 5 - 6 843 4511762 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001719.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Timm21

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          protein transport
          protein import into mitochondrial matrix
          mitochondrial respiratory chain complex I assembly
          mitochondrial cytochrome c oxidase assembly
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31891630 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31891633 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31891632 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395842 BC034297.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395842 AK029037.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395842 AK032468.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395842 AK168105.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395842 NM_025969.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395842 BC034297.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395842 AK029037.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395842 AK032468.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395842 AK168105.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395842 NM_025969.4 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130822 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395842 BC034297.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395842 AK029037.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395842 AK032468.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395842 AK168105.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395842 NM_025969.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395842 BC034297.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395842 AK029037.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395842 AK032468.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395842 AK168105.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395842 NM_025969.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31891634 - -
          Whole Mouse Genome G4122A A_51_P395842 BC034297.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395842 AK029037.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395842 AK032468.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395842 AK168105.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395842 NM_025969.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395842 BC034297.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395842 AK029037.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395842 AK032468.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395842 AK168105.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395842 NM_025969.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102905 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13918711 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13918727 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13918721 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13918730 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13918731 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13918712 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13918728 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31891630 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31891633 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31891632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31891647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13918730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13614945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14018975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13918728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13659042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13825436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13614944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13918721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14018986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13765931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31891656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13614950 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10608682 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460104 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460105 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460103 - -
          Affymetrix HT MG-430 PM Array Plate 1452785_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10460102 - -
          Affymetrix Mouse Gene 1.1 ST Array 10608682 - -
          Mouse Genome 430 2.0 Array 1452785_at - -
          Mouse Genome 430 2.0 Array 1452785_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67105_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67105_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800012226.mm.1 TC1800001719.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800012226.mm.1 NM_025969.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800012226.mm.1 NM_025969.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1800012227.mm.1 TC1800001719.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800012227.mm.1 NM_025969.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800012227.mm.1 NM_025969.4 Refseq
          Mouse Expression Set 430 1452785_at - -
          Mouse Expression Set 430 1452785_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1452785_at - -
          Affymetrix HT Mouse Genome 430B Array 1452785_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6860026 - -
          MouseWG-6 v2 Expression BeadChip 840021 - -
          Illumina mouse-6 v1.0 expression beadchip 6860026 - -
          Illumina mouse-6 v1.0 expression beadchip 840021 - -
          Illumina mouse-6 v1.0 expression beadchip 6100348 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006100348 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000840021 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006860026 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2685133 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2732877 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2684388 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2685133 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2732877 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2684388 - -
          mouse-6 v1.1 (Illumina) scl067105.6_6-S - -
          mouse-6 v1.1 (Illumina) scl067105.1_68-S - -
          mouse-6 v1.1 (Illumina) scl0067105.1_17-S - -
          Illumina Sentrix mouse-6.v1 bead array 6100348 - -
          Illumina Sentrix mouse-6.v1 bead array 6860026 - -
          Illumina Sentrix mouse-6.v1 bead array 840021 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067105.6_6-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0067105.1_17-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067105.1_68-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 840021 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6860026 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6100348 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2684388 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2897476 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2732877 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2685133 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2897476 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2684388 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2732877 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2685133 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067105.6_6-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067105.1_68-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0