Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01264019_m1
          See other NOC2L GE Assays ›
          Gene Symbol:
          Noc2l
          Gene Name:
          nucleolar complex associated 2 homolog (S. cerevisiae)
          Gene Aliases:
          AA410003, AF155546, NIR
          Chromosome Location:
          Chr.4: 156235823 - 156247616 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01264019_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Noc2l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021303.2 NP_067278.2 17 - 18 2093 132
          GenBank mRNA AF155546.1 - 5 - 6 603 132
          AK076287.1 - 16 - 17 2135 132
          AK133834.1 - 17 - 18 2093 132
          AK157765.1 - 17 - 18 2086 132
          AK199140.1 - 1 - 2 96 132
          BC020013.1 - 17 - 18 2064 3663482 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002110.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Noc2l

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          negative regulation of B cell apoptotic process
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          apoptotic process
          chromatin assembly
          cellular response to UV
          negative regulation of histone acetylation
          protein localization to nucleoplasm
          negative regulation of intrinsic apoptotic signaling pathway
          p53 binding
          chromatin binding
          transcription corepressor activity
          RNA binding
          nucleosome binding
          nucleosomal histone binding
          histone binding
          DNA-binding transcription factor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169920 BC026511.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169920 BC026991.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169920 AY458844.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169920 AK161035.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169920 NM_001110516.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169920 BC026511.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169920 BC026991.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169920 AY458844.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169920 AK161035.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169920 NM_001110516.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169920 BC026511.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169920 BC026991.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169920 AY458844.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169920 AK161035.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169920 NM_001110516.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169920 BC026511.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169920 BC026991.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169920 AY458844.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169920 AK161035.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169920 NM_001110516.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169920 BC026511.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169920 BC026991.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169920 AY458844.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169920 AK161035.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169920 NM_001110516.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169920 BC026511.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169920 BC026991.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169920 AY458844.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169920 AK161035.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169920 NM_001110516.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169920 BC026511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169920 BC026991.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169920 AY458844.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169920 AK161035.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169920 NM_001110516.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169920 BC026511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169920 BC026991.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169920 AY458844.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169920 AK161035.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169920 NM_001110516.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P497193 BC020013.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P497193 AK133834.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P497193 AK157765.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P497193 AK076287.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P497193 AK078560.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115617 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10608699 - -
          Affymetrix HT MG-430 PM Array Plate 1436136_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1424323_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10608699 - -
          Mouse Genome 430 2.0 Array 1424323_at - -
          Mouse Genome 430 2.0 Array 1436136_PM_at - -
          Mouse Genome 430 2.0 Array 1424323_PM_at - -
          Mouse Genome 430 2.0 Array 1436136_at - -
          Affymetrix Mouse Clariom S Array PSR0400017840.mm.1 TC0400002110.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400017840.mm.1 NM_021303.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400017840.mm.1 NM_021303.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400017839.mm.1 TC0400002110.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400017839.mm.1 NM_021303.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400017839.mm.1 NM_021303.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400017841.mm.1 TC0400002110.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400017841.mm.1 NM_021303.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400017841.mm.1 NM_021303.2 Refseq
          Mouse Expression Set 430 1424323_PM_at - -
          Mouse Expression Set 430 1424323_at - -
          Mouse Expression Set 430 1436136_PM_at - -
          Mouse Expression Set 430 1436136_at - -
          Mouse Genome 430A 2.0 Array 1424323_PM_at - -
          Mouse Genome 430A 2.0 Array 1424323_at - -
          Affymetrix HT Mouse Genome 430A Array 1424323_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424323_at - -
          Affymetrix HT Mouse Genome 430B Array 1436136_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436136_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5290035 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005290035 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2608162 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2608162 - -
          mouse-6 v1.1 (Illumina) scl24592.15.1_12-S - -
          Illumina Sentrix mouse-6.v1 bead array 5290035 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24592.15.1_12-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5290035 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2851005 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl24592.15.1_12-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0