Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01264396_m1
          See other CSNK1D GE Assays ›
          Gene Symbol:
          Csnk1d
          Gene Name:
          casein kinase 1, delta
          Gene Aliases:
          1200006A05Rik, AA409348, D930010H05Rik
          Chromosome Location:
          Chr.11: 120958010 - 120991333 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01264396_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csnk1d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027874.2 NP_082150.1 1 - 2 400 74
          NM_139059.2 NP_620690.1 1 - 2 400 74
          GenBank mRNA AK004606.1 - 1 - 2 160 74
          AK051052.1 - 1 - 2 399 74
          AK088642.1 - 1 - 2 135 74
          AK140317.1 - 1 - 2 125 74
          AK143534.1 - 1 - 2 100 74
          AK146600.1 - 1 - 2 390 74
          AK150815.1 - 1 - 2 127 74
          AK152721.1 - 1 - 2 342 74
          AK157812.1 - 1 - 2 135 74
          AK163584.1 - 1 - 2 119 74
          BC004604.1 - 1 - 2 386 3585286 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100004236.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Csnk1d

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          protein phosphorylation
          endocytosis
          regulation of cell shape
          Wnt signaling pathway
          phosphorylation
          peptidyl-serine phosphorylation
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          circadian regulation of gene expression
          regulation of circadian rhythm
          rhythmic process
          spindle assembly
          positive regulation of canonical Wnt signaling pathway
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          peptide binding
          tau-protein kinase activity
          phosphoprotein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100040635.mm.1 TC1100004236.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040635.mm.1 NM_027874.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040635.mm.1 NM_027874.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040635.mm.1 NM_139059.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040635.mm.1 NM_139059.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040643.mm.1 TC1100004236.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040643.mm.1 NM_027874.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040643.mm.1 NM_027874.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040643.mm.1 NM_139059.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040643.mm.1 NM_139059.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040638.mm.1 TC1100004236.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040638.mm.1 NM_027874.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040638.mm.1 NM_027874.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040638.mm.1 NM_139059.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040638.mm.1 NM_139059.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040640.mm.1 TC1100004236.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040640.mm.1 NM_027874.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040640.mm.1 NM_027874.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040640.mm.1 NM_139059.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040640.mm.1 NM_139059.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040641.mm.1 TC1100004236.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040641.mm.1 NM_027874.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040641.mm.1 NM_027874.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040641.mm.1 NM_139059.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040641.mm.1 NM_139059.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040642.mm.1 TC1100004236.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040642.mm.1 NM_027874.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040642.mm.1 NM_027874.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040642.mm.1 NM_139059.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040642.mm.1 NM_139059.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 852430 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0