Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01264809_g1
          See other ST8SIA3OS GE Assays ›
          Gene Symbol:
          St8sia3os
          Gene Name:
          ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3, opposite strand
          Gene Aliases:
          1700091E21Rik
          Chromosome Location:
          Chr.18: 64157724 - 64277180 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01264809_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: St8sia3os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045366.1 5 - 6 546 99
          GenBank mRNA BC048666.1 - 5 - 6 546 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream WD repeat domain 7 Wdr7 AI462727,
          TRAG,
          mKIAA0541
          -168.0kb Anti-sense Chr. 18 - 63708695 - 63989760 on NCBI Build GRCm38
          Downstream ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3 St8sia3 AI847333,
          ST8SiaIII,
          Siat8c
          96.1kb Anti-sense Chr. 18 - 64254359 - 64276144 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001456.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 877668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 730460 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 458378 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 801525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 180583 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 362111 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278585 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 862279 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 645879 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 929481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 761835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 771620 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 698559 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 652468 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 639869 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 846918 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848744 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848863 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 157163 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813776 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365433 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837706 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 563862 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394348 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 828919 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 945908 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 504419 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48778 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593206 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29485 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354546 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197291 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 147679 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 112265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55066 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 27754 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86472 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 98449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13040 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 26865 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150287 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150287 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03496314 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09028776 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09028729 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09028781 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09028811 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09028682 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09028827 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09028874 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09028861 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09028893 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110098 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09028874 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09028781 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150287 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03496314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13912008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13774703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13772907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14021766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13604072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13604081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14061034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14061035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14042997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13604070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13912007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13604083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13604071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13856143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14135844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13772936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13717634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13772933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13717637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13946234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13604077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13717635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13972881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14043002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31774039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13856138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31773916 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800010718.mm.1 TC1800001456.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800010718.mm.1 NR_045366.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010718.mm.1 NR_045366.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 368229 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104010309 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0