Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01264828_m1
          See other 1700065O20RIK GE Assays ›
          Gene Symbol:
          1700065O20Rik
          Gene Name:
          RIKEN cDNA 1700065O20 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 49803332 - 49817956 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01264828_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700065O20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045386.1 4 - 5 513 64
          GenBank mRNA AK006898.1 - 4 - 5 516 64
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY706891.1 - 4 - 5 515 Chr. 18 - 49803326 - 49817958 [-] on Build GRCm38 64

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 16283 Gm16283 OTTMUSG00000033143 -41.8kb Anti-sense Chr. 18 - 49755890 - 49761581 on NCBI Build GRCm38
          Downstream Dmx-like 1 Dmxl1 C630007L23Rik,
          D030030C22
          27.1kb Anti-sense Chr. 18 - 49832993 - 49965473 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001320.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54069 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 680645 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228763 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837637 AK045466.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 894942 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 659169 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 967195 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 18975 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163790 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163790 - -
          Whole Mouse Genome G4122A A_52_P818653 AK006898.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130245 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13816928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13809658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13712140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13743238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14139429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13758754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13907688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14042244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31694807 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800009772.mm.1 TC1800001320.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009772.mm.1 NR_045386.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009772.mm.1 NR_045386.1 Refseq
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102340368 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0