Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01265036_m1
          See other GRIN2B GE Assays ›
          Gene Symbol:
          Grin2b
          Gene Name:
          glutamate receptor, ionotropic, NMDA2B (epsilon 2)
          Gene Aliases:
          AW490526, GluN2B, NR2B, Nmdar2b
          Chromosome Location:
          Chr.6: 135715272 - 136173819 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01265036_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grin2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008171.3 NP_032197.3 10 - 11 2544 60
          GenBank mRNA D10651.2 - 7 - 8 1781 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003321.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Grin2b

          Gene Ontology Categories:

          Function(s) Process(es)

          action potential
          behavioral fear response
          in utero embryonic development
          startle response
          suckling behavior
          transport
          ion transport
          cation transport
          calcium ion transport
          chemical synaptic transmission
          sensory organ development
          learning or memory
          learning
          memory
          associative learning
          embryo development
          regulation of protein kinase A signaling
          positive regulation of cell death
          positive regulation of glutamate secretion
          ion transmembrane transport
          ionotropic glutamate receptor signaling pathway
          synaptic transmission, glutamatergic
          fear response
          receptor clustering
          regulation of MAPK cascade
          response to ethanol
          regulation of synaptic plasticity
          regulation of neuronal synaptic plasticity
          regulation of long-term neuronal synaptic plasticity
          behavioral response to pain
          rhythmic process
          positive regulation of synaptic transmission
          detection of mechanical stimulus involved in sensory perception of pain
          regulation of postsynaptic membrane potential
          excitatory postsynaptic potential
          long-term synaptic potentiation
          calcium ion transmembrane transport
          cation transmembrane transport
          ionotropic glutamate receptor activity
          NMDA glutamate receptor activity
          signaling receptor binding
          interleukin-1 receptor binding
          ion channel activity
          extracellularly glutamate-gated ion channel activity
          cation channel activity
          calcium channel activity
          protein binding
          beta-catenin binding
          zinc ion binding
          glycine binding
          D2 dopamine receptor binding
          ionotropic glutamate receptor binding
          signaling receptor activity
          neurotransmitter binding
          metal ion binding
          protein heterodimerization activity
          cell adhesion molecule binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600026036.mm.1 TC0600003321.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600026036.mm.1 NM_008171.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026036.mm.1 NM_008171.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0