Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01265111_m1
          See other INO80DOS GE Assays ›
          Gene Symbol:
          Ino80dos
          Gene Name:
          INO80 complex subunit D, opposite strand
          Gene Aliases:
          Gm11602, OTTMUSG00000002611
          Chromosome Location:
          Chr.1: 63114471 - 63158295 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01265111_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ino80dos
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045914.1 3 - 4 388 195
          NR_045915.1 3 - 4 241 195
          GenBank mRNA AK017924.1 - 3 - 4 241 195
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CK618004.1 - 3 - 4 353 Chr. 1 - 63114775 - 63157127 [+] on Build GRCm38 196
          GenBankmRNA BY717785.1 - 3 - 4 241 Chr. 1 - 63114471 - 63157305 [+] on Build GRCm38 187
          GenBankmRNA DN174372.1 - 3 - 4 394 Chr. 1 - 63114750 - 63157245 [+] on Build GRCm38 195

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream NADH dehydrogenase (ubiquinone) Fe-S protein 1 Ndufs1 5830412M15Rik,
          9930026A05Rik
          Anti-sense Chr. 1 - 63143592 - 63176822 on NCBI Build GRCm38
          Downstream eukaryotic translation elongation factor 1 beta 2 Eef1b2 2810017J07Rik,
          Eef1b
          19.5kb Sense Chr. 1 - 63176831 - 63180486 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000517.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20988 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 538293 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 243410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 246000 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 486474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724230 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48329 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409203 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 49436 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 348702 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 167497 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286748 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198282 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 261505 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471235 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 51631 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706827 AJ009781.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706827 AK003363.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706827 BC103790.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706827 NM_053088.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84703 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 145796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12385 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35416 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273817 AK017924.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04121591 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04121556 - -
          Whole Mouse Genome G4122A A_51_P273817 AK017924.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P20285285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20285292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00458511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20285289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00850940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20285294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20285295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20285290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20285286 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453354_PM_at - -
          Mouse Genome 430 2.0 Array 1453354_at - -
          Mouse Genome 430 2.0 Array 1453354_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100004140.mm.1 TC0100000517.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100004140.mm.1 NR_045914.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100004140.mm.1 NR_045914.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100004140.mm.1 NR_045915.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100004140.mm.1 NR_045915.1 Refseq
          Mouse Expression Set 430 1453354_at - -
          Mouse Expression Set 430 1453354_PM_at - -
          Murine Genome U74v2 Set 139980_g_at - -
          Murine Genome U74v2 Set 139979_PM_at - -
          Murine Genome U74v2 Set 139980_PM_g_at - -
          Murine Genome U74v2 Set 139979_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 777779 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0