Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01265161_g1
          See other GM5095 GE Assays ›
          Gene Symbol:
          Gm5095
          Gene Name:
          predicted gene 5095
          Gene Aliases:
          A730092B10
          Chromosome Location:
          Chr.18: 47537735 - 47718636 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01265161_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm5095
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033454.1 5 - 6 1473 97
          GenBank mRNA AK043399.1 - 5 - 6 1474 97
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB259914.2 - 5 - 6 630 Chr. 18 - 47538578 - 47718633 [+] on Build GRCm38 97

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 9130209A04 gene 9130209A04Rik -247.5kb Sense Chr. 18 - 47452668 - 47470868 on NCBI Build GRCm38
          Downstream predicted gene 4146 Gm4146 42.5kb Anti-sense Chr. 18 - 47761088 - 47789533 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000496.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 469542 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 862338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 606937 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 385414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140938 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810621 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596698 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 159637 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63439 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926618 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367285 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 817346 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 889914 AK080421.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 889914 AK081953.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 889914 AK155011.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 889914 BC026578.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 404855 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813451 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 221132 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 715835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824211 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187300 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 857988 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733880 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 552984 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 268541 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 234927 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290375 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 72160 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320197 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 150630 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313917 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906724 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 304462 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923242 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 106882 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 170331 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409229 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 959749 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 317260 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 951795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 663746 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 933547 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187368 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 849978 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 253531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464962 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529974 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 486964 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211019 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 51903 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 220640 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 959665 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 281476 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 644494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812408 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 603633 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 718326 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115131 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487019 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 77059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 29567 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 72260 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51530 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 93876 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 24725 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13120 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58363 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 161889 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 50803 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P130840 AK043399.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P130840 NR_033454.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08991391 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08991056 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08991331 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08991315 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08991099 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08991217 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03469084 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08991366 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08991275 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08991243 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08991179 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08991131 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03469118 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08991153 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03468841 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123770 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130840 AK043399.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130840 NR_033454.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08991217 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03469084 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03468841 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08991366 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03469118 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08991275 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08991056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31684063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13634547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13701187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13760596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13927908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13819400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13823866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14087185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13701490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31684074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13823861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13801526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31684071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13889514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13898370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31684059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13897021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13889505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13653073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31684079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13819402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31684078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31684072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13766160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31684068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14002033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14095337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14132889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31684075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13760591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31684060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13986277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31684080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31684066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31684062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13801530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14132893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13892833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13760595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14095341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31684082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31683529 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800003583.mm.1 TC1800000496.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800003583.mm.1 NR_033454.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800003583.mm.1 NR_033454.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 340275 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline