Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01265987_g1
          See other SS18 GE Assays ›
          Gene Symbol:
          Ss18
          Gene Name:
          synovial sarcoma translocation, Chromosome 18
          Gene Aliases:
          D130059H17, Ssxt, Syt
          Chromosome Location:
          Chr.18: 14625629 - 14683002 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01265987_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ss18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001161369.1 NP_001154841.1 3 - 4 415 129
          NM_001161370.1 NP_001154842.1 3 - 4 415 129
          NM_001161371.1 NP_001154843.1 3 - 4 415 129
          NM_009280.2 NP_033306.2 3 - 4 415 129
          GenBank mRNA AK039430.1 - 3 - 4 265 129
          AK051601.1 - 268 129
          AK052476.1 - 3 - 4 269 129
          AK083622.1 - 3 - 4 277 129
          AK083887.1 - 3 - 4 299 129
          AK149325.1 - 3 - 4 277 129
          AK166053.1 - 3 - 4 268 129
          BC065062.1 - 3 - 4 252 129
          BC096742.1 - 3 - 4 252 6416415 129
          CF104258.1 - 3 - 4 413 129
          X93357.1 - 3 - 4 415 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001009.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ss18

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          cell morphogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cytoskeleton organization
          response to xenobiotic stimulus
          intracellular signal transduction
          positive regulation of transcription by RNA polymerase II
          ephrin receptor signaling pathway
          neuronal stem cell population maintenance
          nuclear receptor coactivator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800007541.mm.1 TC1800001009.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800007541.mm.1 NM_001161369.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007541.mm.1 NM_001161369.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007541.mm.1 NM_009280.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007541.mm.1 NM_009280.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007542.mm.1 TC1800001009.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800007542.mm.1 NM_001161369.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007542.mm.1 NM_001161369.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007542.mm.1 NM_001161370.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007542.mm.1 NM_001161370.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007542.mm.1 NM_001161371.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007542.mm.1 NM_001161371.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007542.mm.1 NM_009280.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007542.mm.1 NM_009280.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007540.mm.1 TC1800001009.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800007540.mm.1 NM_001161369.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007540.mm.1 NM_001161369.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007540.mm.1 NM_001161370.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007540.mm.1 NM_001161370.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007540.mm.1 NM_001161371.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007540.mm.1 NM_001161371.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007540.mm.1 NM_009280.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007540.mm.1 NM_009280.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0