Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01266654_g1
          See other CSF1R GE Assays ›
          Gene Symbol:
          Csf1r
          Gene Name:
          colony stimulating factor 1 receptor
          Gene Aliases:
          AI323359, CD115, CSF-1R, Csfmr, Fim-2, Fms, M-CSF-R, M-CSFR
          Chromosome Location:
          Chr.18: 61096808 - 61131139 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01266654_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csf1r
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037859.2 NP_001032948.2 19 - 20 2829 108
          GenBank mRNA AK004947.1 - 18 - 19 2639 108
          AK076215.1 - 18 - 19 2638 108
          AK079247.1 - 18 - 19 2646 108
          AK080326.1 - 18 - 19 2627 108
          AK143545.1 - 18 - 19 2630 108
          AK146069.1 - 19 - 20 2830 108
          AK154128.1 - 18 - 19 2644 108
          AK154556.1 - 18 - 19 2640 108
          AK154653.1 - 18 - 19 2640 108
          AK154699.1 - 18 - 19 2640 108
          AK155488.1 - 18 - 19 2650 108
          AK155498.1 - 4 - 5 406 108
          AK155565.1 - 14 - 15 2139 108
          AK155576.1 - 2646 108
          AK155640.1 - 18 - 19 2644 108
          AK155654.1 - 18 - 19 2644 108
          AK155858.1 - 18 - 19 2647 108
          AK156550.1 - 18 - 19 2664 108
          AK171386.1 - 18 - 19 2644 108
          BC036343.1 - 18 - 19 2616 4948135 108
          BC043054.1 - 18 - 19 2620 6418718 108
          BC050024.1 - 18 - 19 2727 108
          BC066863.1 - 19 - 20 2738 30436119 108
          X06368.1 - 18 - 19 2619 108
          X68932.1 - 18 - 19 2529 108

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Csf1r

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          immune system process
          protein phosphorylation
          inflammatory response
          transmembrane receptor protein tyrosine kinase signaling pathway
          cell population proliferation
          positive regulation of cell population proliferation
          regulation of cell shape
          phosphorylation
          peptidyl-tyrosine phosphorylation
          cytokine-mediated signaling pathway
          hemopoiesis
          osteoclast differentiation
          positive regulation of cell migration
          ruffle organization
          positive regulation of chemokine production
          cellular response to macrophage colony-stimulating factor stimulus
          macrophage colony-stimulating factor signaling pathway
          positive regulation of tyrosine phosphorylation of STAT protein
          innate immune response
          regulation of bone resorption
          cell-cell junction maintenance
          positive regulation of osteoclast differentiation
          phosphatidylinositol metabolic process
          protein autophosphorylation
          phosphatidylinositol-mediated signaling
          positive regulation of protein tyrosine kinase activity
          positive regulation of ERK1 and ERK2 cascade
          cellular response to cytokine stimulus
          positive regulation of protein serine/threonine kinase activity
          positive regulation of cell motility
          regulation of actin cytoskeleton reorganization
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          macrophage colony-stimulating factor receptor activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein phosphatase binding
          cytokine binding
          protein homodimerization activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0