Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01267425_m1
          See other 2700046A07RIK GE Assays ›
          Gene Symbol:
          2700046A07Rik
          Gene Name:
          RIKEN cDNA 2700046A07 gene
          Gene Aliases:
          A330042I05Rik
          Chromosome Location:
          Chr.18: 62751674 - 62756347 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01267425_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2700046A07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037693.1 2 - 3 475 70
          GenBank mRNA AK012381.1 - 2 - 3 404 70
          BC072581.1 - 2 - 3 274 70
          CJ067024.1 - 2 - 3 444 70
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CD553290.1 - 2 - 3 74 Chr. 18 - 62752831 - 62755946 [-] on Build GRCm38 70

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream serine peptidase inhibitor,
          Kazal type 13
          Spink13 ENSMUSG00000073551,
          Gm10534,
          Spink5l3
          -10.3kb Sense Chr. 18 - 62605898 - 62741387 on NCBI Build GRCm38
          Downstream adenomatosis polyposis coli down-regulated 1 Apcdd1 AB023957,
          AU041258,
          Drapc1,
          EIG180
          167.2kb Anti-sense Chr. 18 - 62922327 - 62953195 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001445.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371892 BC072581.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371892 NR_037693.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371892 AK012381.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371892 BC072581.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371892 NR_037693.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371892 AK012381.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2143572 AK020725.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126248 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371892 BC072581.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371892 NR_037693.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371892 AK012381.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371892 BC072581.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371892 NR_037693.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371892 AK012381.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371892 BC072581.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371892 NR_037693.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371892 AK012381.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371892 BC072581.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371892 NR_037693.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371892 AK012381.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P241751 AK020725.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13723507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13647536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13642389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13782308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13645424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13647537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13681939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13805835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13782305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13805837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13696024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14034667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13782311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13814657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31765407 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10459334 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459333 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459330 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459332 - -
          Affymetrix HT MG-430 PM Array Plate 1453645_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1430118_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10459329 - -
          Mouse Genome 430 2.0 Array 1430118_at - -
          Mouse Genome 430 2.0 Array 1453645_at - -
          Mouse Genome 430 2.0 Array 1430118_PM_at - -
          Mouse Genome 430 2.0 Array 1453645_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78449_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78449_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800010617.mm.1 TC1800001445.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800010617.mm.1 NR_037693.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010617.mm.1 NR_037693.1 Refseq
          Mouse Expression Set 430 1430118_PM_at - -
          Mouse Expression Set 430 1453645_PM_at - -
          Mouse Expression Set 430 1453645_at - -
          Mouse Expression Set 430 1430118_at - -
          Murine Genome U74v2 Set 169420_PM_at - -
          Murine Genome U74v2 Set 169420_at - -
          Affymetrix HT Mouse Genome 430B Array 1430118_at - -
          Affymetrix HT Mouse Genome 430B Array 1453645_at - -
          Affymetrix HT Mouse Genome 430B Array 1430118_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453645_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 852722 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101740022 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214052 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0