Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01267938_m1
          See other 1700060C16RIK GE Assays ›
          Gene Symbol:
          1700060C16Rik
          Gene Name:
          RIKEN cDNA 1700060C16 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 143593353 - 143651288 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01267938_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700060C16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045732.1 4 - 5 463 110
          GenBank mRNA AK006838.1 - 4 - 5 463 110

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4930579D09 gene 4930579D09Rik -57.8kb Sense Chr. 6 - 143533943 - 143535533 on NCBI Build GRCm38
          Downstream SRY (sex determining region Y)-box 5 Sox5 A730017D01Rik,
          AI528773
          214.5kb Sense Chr. 6 - 143828425 - 144781989 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003396.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 468200 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806631 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 759652 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 548886 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 816365 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 94434 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 311644 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 538373 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 321226 AK085018.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 602956 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 64340 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 870267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52906 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 14192 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102905 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01464822 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06097602 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06097629 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06097519 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01464842 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06097602 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06097519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05056423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05244082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05680037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05330344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05702803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05330331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05244074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05194301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05194292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05443241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05056432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05011037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05286715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05522262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05330345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05056428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05769926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05194297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05244080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05406848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05164652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05769935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05330339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05680039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05867958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05330341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05687430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05056424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05167125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05194291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05194300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05167122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24911385 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600026762.mm.1 TC0600003396.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600026762.mm.1 NR_045732.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026762.mm.1 NR_045732.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 359959 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0