Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01268275_m1
          See other EHD2 GE Assays ›
          Gene Symbol:
          Ehd2
          Gene Name:
          EH-domain containing 2
          Gene Aliases:
          BC027084, C130052H20Rik
          Chromosome Location:
          Chr.7: 15946966 - 15967535 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01268275_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ehd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153068.3 NP_694708.2 5 - 6 1250 63
          GenBank mRNA AK029240.1 - 5 - 6 1195 63
          AK031203.1 - 5 - 6 1252 63
          AK046566.1 - 5 - 6 1192 63
          AK048356.1 - 5 - 6 1195 63
          AK050243.1 - 5 - 6 1230 63
          AK084455.1 - 5 - 6 1193 63
          AK085016.1 - 5 - 6 1193 63
          AY531389.1 - 5 - 6 1189 63
          BC027084.1 - 6 - 7 897 5356224 63
          BC031731.1 - 1115 63
          BC113161.1 - 6 - 7 1208 40060184 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002417.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ehd2

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          actin cytoskeleton organization
          cortical actin cytoskeleton organization
          endocytic recycling
          protein localization to plasma membrane
          positive regulation of myoblast fusion
          positive regulation of endocytic recycling
          nucleotide binding
          calcium ion binding
          protein binding
          ATP binding
          GTP binding
          hydrolase activity
          protein domain specific binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343613 BC113161.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343613 BC027084.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343613 AK085016.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343613 AY531389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343613 AK050243.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343613 BC113161.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343613 BC027084.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343613 AK085016.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343613 AY531389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343613 AK050243.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076906 AK029557.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076906 AK029240.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573715 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 382662 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 382680 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 656173 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 644314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 24177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181889 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 684879 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 245813 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284972 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360581 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 5078 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115317 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135064 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127311 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343613 BC113161.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343613 BC027084.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343613 AK085016.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343613 AY531389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343613 AK050243.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343613 BC113161.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343613 BC027084.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343613 AK085016.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343613 AY531389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343613 AK050243.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076906 AK029557.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076906 AK029240.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P129430 BC031731.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P129430 BC020014.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P343613 BC113161.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P343613 BC027084.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P343613 AK085016.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P343613 AY531389.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P343613 AK050243.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P343613 BC113161.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P343613 BC027084.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P343613 AK085016.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P343613 AY531389.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P343613 AK050243.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03807736 - -
          Whole Mouse Genome G4122A A_52_P340595 - -
          Whole Mouse Genome G4122A A_51_P343613 BC113161.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343613 BC027084.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343613 AK085016.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343613 AY531389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343613 AK050243.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343613 BC113161.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343613 BC027084.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343613 AK085016.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343613 AY531389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343613 AK050243.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P129430 BC031731.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P129430 BC020014.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123076 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24978925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24978910 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560193 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560192 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560201 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560200 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560197 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560199 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560194 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560195 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560198 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560196 - -
          Affymetrix HT MG-430 PM Array Plate 1424157_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1424158_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1459823_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456166_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1427729_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1435785_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10560190 - -
          Mouse Genome 430 2.0 Array 1424157_at - -
          Mouse Genome 430 2.0 Array 1456166_PM_at - -
          Mouse Genome 430 2.0 Array 1424157_PM_at - -
          Mouse Genome 430 2.0 Array 1456166_at - -
          Mouse Genome 430 2.0 Array 1424158_at - -
          Mouse Genome 430 2.0 Array 1424158_PM_at - -
          Mouse Genome 430 2.0 Array 1427729_PM_at - -
          Mouse Genome 430 2.0 Array 1427729_at - -
          Mouse Genome 430 2.0 Array 1435785_PM_at - -
          Mouse Genome 430 2.0 Array 1435785_at - -
          Mouse Genome 430 2.0 Array 1459823_at - -
          Mouse Genome 430 2.0 Array 1459823_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 259300_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 259300_at - -
          Affymetrix Mouse Clariom S Array PSR0700021244.mm.1 TC0700002417.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021244.mm.1 NM_153068.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021244.mm.1 NM_153068.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021243.mm.1 TC0700002417.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021243.mm.1 NM_153068.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021243.mm.1 NM_153068.3 Refseq
          Mouse Expression Set 430 1427729_PM_at - -
          Mouse Expression Set 430 1427729_at - -
          Mouse Expression Set 430 1424158_at - -
          Mouse Expression Set 430 1424157_PM_at - -
          Mouse Expression Set 430 1424158_PM_at - -
          Mouse Expression Set 430 1424157_at - -
          Mouse Expression Set 430 1459823_PM_at - -
          Mouse Expression Set 430 1456166_PM_at - -
          Mouse Expression Set 430 1435785_PM_at - -
          Mouse Expression Set 430 1459823_at - -
          Mouse Expression Set 430 1435785_at - -
          Mouse Expression Set 430 1456166_at - -
          Murine Genome U74v2 Set 166065_f_at - -
          Murine Genome U74v2 Set 166065_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1424157_at - -
          Mouse Genome 430A 2.0 Array 1424158_at - -
          Mouse Genome 430A 2.0 Array 1424158_PM_at - -
          Mouse Genome 430A 2.0 Array 1427729_PM_at - -
          Mouse Genome 430A 2.0 Array 1424157_PM_at - -
          Mouse Genome 430A 2.0 Array 1427729_at - -
          Affymetrix HT Mouse Genome 430A Array 1424158_at - -
          Affymetrix HT Mouse Genome 430A Array 1424157_at - -
          Affymetrix HT Mouse Genome 430A Array 1424157_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427729_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427729_at - -
          Affymetrix HT Mouse Genome 430A Array 1424158_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456166_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435785_at - -
          Affymetrix HT Mouse Genome 430B Array 1435785_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459823_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456166_at - -
          Affymetrix HT Mouse Genome 430B Array 1459823_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106400048 - -
          Illumina mouse-6 v1.0 expression beadchip 6040044 - -
          Illumina mouse-6 v1.0 expression beadchip 106400048 - -
          Illumina mouse-6 v1.0 expression beadchip 106400520 - -
          Illumina mouse-6 v1.0 expression beadchip 102030082 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006040044 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2698921 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229953 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255622 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2698921 - -
          mouse-6 v1.1 (Illumina) scl36563.1.1_32-S - -
          Illumina Sentrix mouse-6.v1 bead array 6040044 - -
          Illumina Sentrix mouse-6.v1 bead array 102030082 - -
          Illumina Sentrix mouse-6.v1 bead array 106400520 - -
          Illumina Sentrix mouse-6.v1 bead array 106400048 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36563.1.1_32-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106400520 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6040044 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102030082 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106400048 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2810645 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2698921 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2557496 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2554000 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255622 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2810645 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0