Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01268944_mH
          See other SEC23B GE Assays ›
          Gene Symbol:
          Sec23b
          Gene Name:
          SEC23B (S. cerevisiae)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 144556229 - 144590753 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01268944_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sec23b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001252543.1 NP_001239472.1 14 - 15 1787 115
          NM_001252544.1 NP_001239473.1 14 - 15 1780 115
          NM_001252545.1 NP_001239474.1 14 - 15 1769 115
          NM_001281816.1 NP_001268745.1 11 - 12 1409 115
          NM_019787.4 NP_062761.2 14 - 15 1798 115
          GenBank mRNA AF200326.1 - 13 - 14 1670 115
          AK014597.1 - 14 - 15 1748 115
          AK029661.1 - 11 - 12 1410 115
          AK139729.1 - 14 - 15 1713 115
          AK146083.1 - 14 - 15 1747 115
          AK149693.1 - 14 - 15 1727 115
          AK153214.1 - 14 - 15 1754 115
          AK158398.1 - 14 - 15 1765 115
          AK166330.1 - 14 - 15 1730 115
          AK167382.1 - 14 - 15 1735 115
          AK167732.1 - 14 - 15 1716 115
          AK171597.1 - 14 - 15 1728 115
          BC005464.1 - 14 - 15 1723 3591420 115
          BC011160.1 - 14 - 15 1750 4024353 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002150.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sec23b

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          intracellular protein transport
          endoplasmic reticulum to Golgi vesicle-mediated transport
          protein transport
          vesicle-mediated transport
          protein binding
          zinc ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200016996.mm.1 TC0200002150.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200016996.mm.1 NM_001252543.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016996.mm.1 NM_001252543.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016996.mm.1 NM_001252544.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016996.mm.1 NM_001252544.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016996.mm.1 NM_001252545.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016996.mm.1 NM_001252545.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016996.mm.1 NM_001281816.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016996.mm.1 NM_019787.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016996.mm.1 NM_019787.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016998.mm.1 TC0200002150.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200016998.mm.1 NM_001252543.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016998.mm.1 NM_001252543.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016998.mm.1 NM_001252544.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016998.mm.1 NM_001252544.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016998.mm.1 NM_001252545.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016998.mm.1 NM_001252545.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016998.mm.1 NM_001281816.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016998.mm.1 NM_019787.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016998.mm.1 NM_019787.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016995.mm.1 TC0200002150.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200016995.mm.1 NM_001252543.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016995.mm.1 NM_001252543.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016995.mm.1 NM_001252544.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016995.mm.1 NM_001252544.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016995.mm.1 NM_001252545.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016995.mm.1 NM_001252545.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016995.mm.1 NM_001281816.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016995.mm.1 NM_019787.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016995.mm.1 NM_019787.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0