Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01269063_m1
          See other IFIH1 GE Assays ›
          Gene Symbol:
          Ifih1
          Gene Name:
          interferon induced with helicase C domain 1
          Gene Aliases:
          9130009C22Rik, Helicard, Hlcd, MDA5, RLR-2
          Chromosome Location:
          Chr.2: 62595612 - 62646255 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01269063_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifih1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164477.1 NP_001157949.1 4 - 5 1272 129
          NM_027835.3 NP_082111.2 5 - 6 1419 129
          GenBank mRNA AF374384.1 - 5 - 6 1328 129
          AK037057.1 - 5 - 6 1317 129
          AK040519.1 - 5 - 6 1376 129
          AK153018.1 - 5 - 6 1326 129
          AK168646.1 - 5 - 6 1420 129
          AY075132.1 - 5 - 6 1094 129
          BC080200.1 - 4 - 5 1177 3597815 129
          EF113547.1 - 5 - 6 1108 129
          EF113548.1 - 5 - 6 1108 129
          EU084750.1 - 5 - 6 1108 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003630.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifih1

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          response to virus
          protein sumoylation
          positive regulation of interferon-alpha production
          positive regulation of interferon-beta production
          regulation of apoptotic process
          innate immune response
          defense response to virus
          nucleotide binding
          DNA binding
          RNA binding
          double-stranded RNA binding
          single-stranded RNA binding
          helicase activity
          protein binding
          ATP binding
          zinc ion binding
          hydrolase activity
          hydrolase activity, acting on acid anhydrides
          ribonucleoprotein complex binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200031003.mm.1 TC0200003630.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200031003.mm.1 NM_001164477.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031003.mm.1 NM_001164477.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031003.mm.1 NM_027835.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031003.mm.1 NM_027835.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031002.mm.1 TC0200003630.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200031002.mm.1 NM_001164477.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031002.mm.1 NM_001164477.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031002.mm.1 NM_027835.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031002.mm.1 NM_027835.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-stddt:80/100.66.134.214:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0