Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01269857_m1
          See other ARRB1 GE Assays ›
          Gene Symbol:
          Arrb1
          Gene Name:
          arrestin, beta 1
          Gene Aliases:
          1200006I17Rik, AW208571, G430100A01Rik
          Chromosome Location:
          Chr.7: 99535486 - 99606772 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01269857_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arrb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177231.2 NP_796205.1 3 - 4 408 101
          NM_178220.3 NP_835738.1 3 - 4 408 101
          GenBank mRNA AK052588.1 - 3 - 4 185 101
          AK090090.1 - 3 - 4 235 101
          AK144054.1 - 3 - 4 207 101
          AK147508.1 - 3 - 4 409 101
          AK147566.1 - 3 - 4 429 101
          AK165514.1 - 3 - 4 207 101
          BC108969.1 - 3 - 4 175 40047224 101
          BC108970.1 - 3 - 4 175 40047225 101

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arrb1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          G protein-coupled receptor internalization
          positive regulation of receptor internalization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          transport
          endocytosis
          receptor-mediated endocytosis
          signal transduction
          G protein-coupled receptor signaling pathway
          phototransduction
          regulation of G protein-coupled receptor signaling pathway
          negative regulation of signal transduction
          protein transport
          protein ubiquitination
          negative regulation of protein ubiquitination
          positive regulation of protein ubiquitination
          negative regulation of NF-kappaB transcription factor activity
          positive regulation of protein binding
          negative regulation of interleukin-6 production
          negative regulation of interleukin-8 production
          positive regulation of peptidyl-serine phosphorylation
          negative regulation of GTPase activity
          positive regulation of smooth muscle cell apoptotic process
          positive regulation of Rho protein signal transduction
          positive regulation of histone acetylation
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          follicle-stimulating hormone signaling pathway
          stress fiber assembly
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of GTPase activity
          positive regulation of transcription by RNA polymerase II
          negative regulation of ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of histone H4 acetylation
          transcription cis-regulatory region binding
          GTPase activator activity
          insulin-like growth factor receptor binding
          protein binding
          transcription factor binding
          estrogen receptor binding
          mitogen-activated protein kinase kinase binding
          ubiquitin protein ligase binding
          alpha-1A adrenergic receptor binding
          alpha-1B adrenergic receptor binding
          angiotensin receptor binding
          follicle-stimulating hormone receptor binding
          V2 vasopressin receptor binding
          AP-2 adaptor complex binding
          clathrin adaptor activity
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          transmembrane transporter binding
          protein phosphorylated amino acid binding
          phosphoprotein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0