Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01271575_g1
          See other RBMS1 GE Assays ›
          Gene Symbol:
          Rbms1
          Gene Name:
          RNA binding motif, single stranded interacting protein 1
          Gene Aliases:
          2600014B10Rik, AI255215, MSSP-1, MSSP-2, MSSP-3, YC1
          Chromosome Location:
          Chr.2: 60750193 - 60963450 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01271575_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbms1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001141931.1 NP_001135403.1 8 - 9 1042 74
          NM_001141932.1 NP_001135404.1 8 - 9 1042 74
          NM_020296.2 NP_064692.2 8 - 9 1235 74
          GenBank mRNA AB026569.1 - 8 - 9 958 74
          AK133054.1 - 8 - 9 763 74
          AK135280.1 - 8 - 9 922 74
          AK145897.1 - 8 - 9 1044 74
          AK147109.1 - 8 - 9 1236 74
          AK161102.1 - 8 - 9 992 74
          AK161416.1 - 4 - 5 372 74
          AK164307.1 - 8 - 9 1036 74
          AK178547.1 - 2 - 3 73 74
          BC016501.1 - 8 - 9 1223 4527742 74
          BC056609.1 - 5 - 6 434 74
          BC057866.1 - 8 - 9 1074 5327143 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003607.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbms1

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          nucleotide binding
          nucleic acid binding
          DNA binding
          RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200030826.mm.1 TC0200003607.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200030826.mm.1 NM_001141931.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030826.mm.1 NM_001141931.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030826.mm.1 NM_001141932.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030826.mm.1 NM_001141932.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030826.mm.1 NM_020296.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030826.mm.1 NM_020296.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030824.mm.1 TC0200003607.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200030824.mm.1 NM_001141932.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030824.mm.1 NM_001141932.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030827.mm.1 TC0200003607.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200030827.mm.1 NM_001141931.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030827.mm.1 NM_001141931.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030827.mm.1 NM_001141932.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030827.mm.1 NM_001141932.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030827.mm.1 NM_020296.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030827.mm.1 NM_020296.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0