Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01273673_m1
          See other NFYA GE Assays ›
          Gene Symbol:
          Nfya
          Gene Name:
          nuclear transcription factor-Y alpha
          Gene Aliases:
          AA407810, Cbf-b, SEZ-10, SEZ10
          Chromosome Location:
          Chr.17: 48386885 - 48409901 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01273673_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nfya
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110832.1 NP_001104302.1 1 - 2 195 102
          NM_010913.2 NP_035043.1 1 - 2 195 102
          GenBank mRNA AK004729.1 - 1 - 2 141 102
          AK137561.1 - 1 - 2 126 102
          AK143723.1 - 1 - 2 157 102
          BC029695.1 - 1 - 2 83 5374597 102
          BC057099.1 - 1 - 2 97 5716111 102
          BY127163.1 - 1 - 2 195 102
          X55315.1 - 1 - 2 105 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002225.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK004729.1, AK143723.1, BC029695.1, BC057099.1, NM_001110832.1, NM_010913.2, X55315.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nfya

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          rhythmic process
          regulation of stem cell population maintenance
          positive regulation of stem cell proliferation
          transcription cis-regulatory region binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          transcription coregulator binding
          DNA binding
          DNA-binding transcription factor activity
          protein binding
          transcription factor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700021049.mm.1 TC1700002225.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700021049.mm.1 NM_001110832.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021049.mm.1 NM_001110832.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021049.mm.1 NM_010913.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021049.mm.1 NM_010913.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021042.mm.1 TC1700002225.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700021042.mm.1 NM_001110832.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021042.mm.1 NM_001110832.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021042.mm.1 NM_010913.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021042.mm.1 NM_010913.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021036.mm.1 TC1700002225.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700021036.mm.1 NM_001110832.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021036.mm.1 NM_001110832.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021037.mm.1 TC1700002225.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700021037.mm.1 NM_001110832.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021037.mm.1 NM_001110832.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021041.mm.1 TC1700002225.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700021041.mm.1 NM_001110832.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021041.mm.1 NM_001110832.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021041.mm.1 NM_010913.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021041.mm.1 NM_010913.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021040.mm.1 TC1700002225.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700021040.mm.1 NM_001110832.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021040.mm.1 NM_001110832.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021040.mm.1 NM_010913.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021040.mm.1 NM_010913.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 671084 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0