Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01273790_g1
          See other PRR3 GE Assays ›
          Gene Symbol:
          Prr3
          Gene Name:
          proline-rich polypeptide 3
          Gene Aliases:
          4930540G07Rik, Cat56
          Chromosome Location:
          Chr.17: 35972539 - 35979825 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01273790_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001282012.1 NP_001268941.1 3 - 4 665 92
          NM_001282013.1 NP_001268942.1 5 - 6 670 92
          NM_001282024.1 NP_001268953.1 4 - 5 764 92
          NM_145487.3 NP_663462.2 3 - 4 693 92
          GenBank mRNA AJ504719.1 - 3 - 4 1462 92
          AK132945.1 - 3 - 4 393 92
          BU148736.1 - 5 - 6 627 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001991.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prr3

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          RNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162042 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162044 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162039 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162046 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162040 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162043 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162041 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2161775 AJ504719.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2161775 NM_145487.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2161775 NR_028507.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2161775 NR_028516.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 166059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 470101 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 893131 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 31641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 84173 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100748 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161775 AJ504719.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161775 NM_145487.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161775 NR_028507.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161775 NR_028516.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162023 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08769738 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100748 - -
          Whole Mouse Genome G4122A A_52_P391825 BP772693.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P391825 AK016009.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P391825 NM_001165892.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498340 BC016636.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109265 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08769738 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100748 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13224912 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13224899 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13224903 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13224914 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13224907 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162041 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162039 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162040 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162044 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162042 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162043 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13285784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13168388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13168389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13089486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13322675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13451307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13451850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13255890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13451852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13451316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13100349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13168383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13255888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13451304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162099 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450671 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450674 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450672 - -
          Affymetrix HT MG-430 PM Array Plate 1424145_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10450669 - -
          Mouse Genome 430 2.0 Array 1424145_at - -
          Mouse Genome 430 2.0 Array 1424145_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75210_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75210_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700019034.mm.1 TC1700001991.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019034.mm.1 NM_001282012.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019034.mm.1 NM_001282013.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019034.mm.1 NM_001282024.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019034.mm.1 NM_145487.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019034.mm.1 NM_145487.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019033.mm.1 TC1700001991.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019033.mm.1 NM_001282012.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019033.mm.1 NM_001282013.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019033.mm.1 NM_001282024.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019033.mm.1 NM_145487.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019033.mm.1 NM_145487.3 Refseq
          Affymetrix HT Mouse Genome 430A Array 1424145_at - -
          Affymetrix HT Mouse Genome 430A Array 1424145_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 436484 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3440112 - -
          Illumina mouse-6 v1.0 expression beadchip 3440112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003440112 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2716365 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2716365 - -
          mouse-6 v1.1 (Illumina) scl49968.5_292-S - -
          Illumina Sentrix mouse-6.v1 bead array 3440112 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49968.5_292-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3440112 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2797322 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2797322 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl49968.5_292-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc