Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01274084_mH
          See other RAB6A GE Assays ›
          Gene Symbol:
          Rab6a
          Gene Name:
          RAB6A, member RAS oncogene family
          Gene Aliases:
          2610028L11Rik, AA419671, Rab6
          Chromosome Location:
          Chr.7: 100607586 - 100641268 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01274084_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab6a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163663.1 NP_001157135.1 1 - 2 707 65
          NM_024287.4 NP_077249.1 1 - 2 707 65
          GenBank mRNA AB041575.1 - 1 - 2 506 65
          AB232596.1 - 1 - 2 69 65
          AK051246.1 - 1 - 2 542 65
          AK083262.1 - 1 - 2 531 65
          AK084131.1 - 1 - 2 515 65
          AK154012.1 - 1 - 2 604 65
          AK160134.1 - 1 - 2 586 65
          AK160866.1 - 1 - 2 515 65
          AK169842.1 - 1 - 2 708 65
          BC019118.1 - 1 - 2 339 3711087 65
          BE134038.1 - 1 - 2 286 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001413.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab6a

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          intracellular protein transport
          retrograde vesicle-mediated transport, Golgi to endoplasmic reticulum
          intra-Golgi vesicle-mediated transport
          small GTPase mediated signal transduction
          protein transport
          vesicle-mediated transport
          peptidyl-cysteine methylation
          antigen processing and presentation
          Rab protein signal transduction
          positive regulation of ATPase activity
          protein localization to Golgi apparatus
          early endosome to Golgi transport
          retrograde transport, endosome to Golgi
          nucleotide binding
          ATPase activator activity
          GTPase activity
          protein binding
          GTP binding
          protein domain specific binding
          myosin V binding
          ATPase binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700012062.mm.1 TC0700001413.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012062.mm.1 NM_001163663.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012062.mm.1 NM_001163663.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012062.mm.1 NM_024287.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012062.mm.1 NM_024287.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012065.mm.1 TC0700001413.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012065.mm.1 NM_001163663.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012065.mm.1 NM_001163663.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012065.mm.1 NM_024287.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012065.mm.1 NM_024287.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012064.mm.1 TC0700001413.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012064.mm.1 NM_001163663.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012064.mm.1 NM_001163663.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012064.mm.1 NM_024287.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012064.mm.1 NM_024287.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012063.mm.1 TC0700001413.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012063.mm.1 NM_001163663.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012063.mm.1 NM_001163663.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012063.mm.1 NM_024287.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012063.mm.1 NM_024287.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012066.mm.1 TC0700001413.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012066.mm.1 NM_001163663.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012066.mm.1 NM_001163663.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012066.mm.1 NM_024287.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012066.mm.1 NM_024287.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 474184 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0