Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01274322_m1
          See other ADK GE Assays ›
          Gene Symbol:
          Adk
          Gene Name:
          adenosine kinase
          Gene Aliases:
          2310026J05Rik, 5033405D03Rik, AI255373, AI987814, Ak
          Chromosome Location:
          Chr.14: 21052573 - 21448569 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01274322_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243041.1 NP_001229970.1 7 - 8 831 125
          NM_134079.4 NP_598840.1 7 - 8 788 125
          GenBank mRNA AK050135.1 - 1 - 2 91 125
          AK151863.1 - 7 - 8 833 125
          AK160264.1 - 7 - 8 785 125
          AY540996.1 - 7 - 8 730 125
          AY540997.1 - 6 - 7 675 125
          BC009659.1 - 7 - 8 753 3485959 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000179.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adk

          Gene Ontology Categories:

          Function(s) Process(es)

          purine ribonucleoside salvage
          AMP biosynthetic process
          dATP biosynthetic process
          positive regulation of cardiac muscle hypertrophy
          phosphorylation
          positive regulation of T cell proliferation
          type B pancreatic cell proliferation
          adenosine metabolic process
          nucleotide binding
          RNA binding
          adenosine kinase activity
          ATP binding
          kinase activity
          transferase activity
          phosphotransferase activity, alcohol group as acceptor
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1400001462.mm.1 TC1400000179.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400001462.mm.1 NM_001243041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001462.mm.1 NM_001243041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001462.mm.1 NM_134079.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001462.mm.1 NM_134079.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001454.mm.1 TC1400000179.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400001454.mm.1 NM_134079.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001454.mm.1 NM_134079.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001461.mm.1 TC1400000179.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400001461.mm.1 NM_001243041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001461.mm.1 NM_001243041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001457.mm.1 TC1400000179.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400001457.mm.1 NM_134079.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001457.mm.1 NM_134079.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001465.mm.1 TC1400000179.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400001465.mm.1 NM_001243041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001465.mm.1 NM_001243041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001465.mm.1 NM_134079.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001465.mm.1 NM_134079.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001463.mm.1 TC1400000179.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400001463.mm.1 NM_001243041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001463.mm.1 NM_001243041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001463.mm.1 NM_134079.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001463.mm.1 NM_134079.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001467.mm.1 TC1400000179.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400001467.mm.1 NM_001243041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001467.mm.1 NM_001243041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001467.mm.1 NM_134079.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001467.mm.1 NM_134079.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001464.mm.1 TC1400000179.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400001464.mm.1 NM_001243041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001464.mm.1 NM_001243041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001464.mm.1 NM_134079.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001464.mm.1 NM_134079.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001466.mm.1 TC1400000179.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400001466.mm.1 NM_001243041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001466.mm.1 NM_001243041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001466.mm.1 NM_134079.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001466.mm.1 NM_134079.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001469.mm.1 TC1400000179.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400001469.mm.1 NM_001243041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001469.mm.1 NM_001243041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001469.mm.1 NM_134079.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001469.mm.1 NM_134079.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 557978 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0