Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01274630_m1
          See other ARL6IP1 GE Assays ›
          Gene Symbol:
          Arl6ip1
          Gene Name:
          ADP-ribosylation factor-like 6 interacting protein 1
          Gene Aliases:
          AIP-6, AL022945, ARMER, AU042858, Aip-1, Arl6ip, C85138, mKIAA0069
          Chromosome Location:
          Chr.7: 118118890 - 118129625 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01274630_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arl6ip1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019419.2 NP_062292.1 1 - 2 156 84
          GenBank mRNA AF133669.1 - 1 - 2 101 84
          AF172088.1 - 1 - 2 142 84
          AF223953.1 - 1 - 2 94 84
          AK090001.1 - 1 - 2 108 84
          AK150042.1 - 1 - 2 109 84
          AK150507.1 - 1 - 2 107 84
          AK151272.1 - 1 - 2 106 84
          AK151507.1 - 1 - 2 106 84
          AK152565.1 - 1 - 2 106 84
          AK152885.1 - 1 - 2 103 84
          AK154355.1 - 1 - 2 128 84
          AK154644.1 - 1 - 2 109 84
          AK163655.1 - 1 - 2 106 84
          AK167872.1 - 1 - 2 111 84
          AK167888.1 - 1 - 2 159 84
          AK167912.1 - 1 - 2 106 84
          AK168172.1 - 1 - 2 109 84
          AK168245.1 - 1 - 2 108 84
          AK169191.1 - 1 - 2 106 84
          BC010196.1 - 1 - 2 96 3157834 84
          BC046792.1 - 1 - 2 79 5717527 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004165.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AF223953.1, AK090001.1, AK150042.1, AK150507.1, AK151272.1, AK151507.1, AK152565.1, AK152885.1, AK154355.1, AK154644.1, AK163655.1, AK167872.1, AK167888.1, AK167912.1, AK168172.1, AK168245.1, AK169191.1, BC010196.1, BC046792.1, NM_019419.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arl6ip1

          Gene Ontology Categories:

          Function(s) Process(es)

          cotranslational protein targeting to membrane
          protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700034560.mm.1 TC0700004165.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700034560.mm.1 NM_019419.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034560.mm.1 NM_019419.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034557.mm.1 TC0700004165.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700034557.mm.1 NM_019419.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034557.mm.1 NM_019419.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034558.mm.1 TC0700004165.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700034558.mm.1 NM_019419.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034558.mm.1 NM_019419.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 931257 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix