Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700018297.mm.1 |
TC0700002055.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018297.mm.1 |
NM_001111050.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018297.mm.1 |
NM_001111050.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018299.mm.1 |
TC0700002055.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018299.mm.1 |
NM_001111049.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018299.mm.1 |
NM_001111049.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018299.mm.1 |
NM_001111050.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018299.mm.1 |
NM_001111050.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018299.mm.1 |
NM_009842.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018299.mm.1 |
NM_009842.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018298.mm.1 |
TC0700002055.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018298.mm.1 |
NM_001111050.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018298.mm.1 |
NM_001111050.1 |
Refseq |