Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01276950_g1
          See other CD163L1 GE Assays ›
          Gene Symbol:
          Cd163l1
          Gene Name:
          CD163 molecule-like 1
          Gene Aliases:
          B430307C05, E430002D04Rik, Scart1
          Chromosome Location:
          Chr.7: 140197218 - 140231145 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01276950_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd163l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172909.4 NP_766497.3 13 - 14 3275 62

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd163l1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989563 AK088010.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989563 AW494409.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989563 AK046675.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989563 NM_172909.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989563 AF539744.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 362717 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 949527 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 619876 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 531028 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 640139 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 684939 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61103 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 95622 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134114 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137225 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989563 AK088010.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989563 AW494409.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989563 AK046675.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989563 NM_172909.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989563 AF539744.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582919 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137225 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134114 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06393905 - -
          Whole Mouse Genome G4122A A_51_P171382 AK088010.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P171382 AW494409.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P171382 AK046675.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P171382 NM_172909.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P171382 AF539744.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101507 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137225 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06195870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06202763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06202770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06202752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06202754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06202766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06204856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06111727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06363999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06202745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06202773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06363995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582933 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10558615 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558628 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558630 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558616 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558625 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558620 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558629 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558621 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558622 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558623 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558624 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558618 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558619 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558617 - -
          Affymetrix HT MG-430 PM Array Plate 1455527_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440808_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10558614 - -
          Mouse Genome 430 2.0 Array 1455527_at - -
          Mouse Genome 430 2.0 Array 1455527_PM_at - -
          Mouse Genome 430 2.0 Array 1440808_x_at - -
          Mouse Genome 430 2.0 Array 1440808_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 244233_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 244233_at - -
          Mouse Expression Set 430 1455527_at - -
          Mouse Expression Set 430 1440808_PM_x_at - -
          Mouse Expression Set 430 1440808_x_at - -
          Mouse Expression Set 430 1455527_PM_at - -
          Murine Genome U74v2 Set 133242_at - -
          Murine Genome U74v2 Set 133242_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440808_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1455527_at - -
          Affymetrix HT Mouse Genome 430B Array 1455527_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440808_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 457910 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1770551 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770551 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1227374 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227374 - -
          mouse-6 v1.1 (Illumina) scl31916.12.1_47-S - -
          Illumina Sentrix mouse-6.v1 bead array 1770551 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31916.12.1_47-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770551 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2629663 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2629663 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0