Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01277526_m1
          See other 5033421B08RIK GE Assays ›
          Gene Symbol:
          5033421B08Rik
          Gene Name:
          RIKEN cDNA 5033421B08 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 8511164 - 8584974 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01277526_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5033421B08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK050516.1 - 2 - 3 350 139

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2303868 AK050516.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199169 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 629002 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493665 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 180133 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 864350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 502348 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 304624 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 338611 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 105414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704514 AK050866.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783817 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79932 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 74932 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112857 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02456721 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02456733 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112857 - -
          Whole Mouse Genome G4122A A_52_P515620 AK050516.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P515620 AK017181.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02456721 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10111693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10407589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09795794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09810108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10279936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10251473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10371115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28223074 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 623985 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100630440 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-64bcc5cc5d-xqgf4:80/100.66.130.62:80.
          git-commit: fd21aaec18e3d7b1379e340bd1771dde3ea5c6fc
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0