Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01277675_m1
          See other 4930543N07RIK GE Assays ›
          Gene Symbol:
          4930543N07Rik
          Gene Name:
          RIKEN cDNA 4930543N07 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 138554242 - 138567643 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01277675_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930543N07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK016032.1 - 4 - 5 772 130

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transcription elongation regulator 1-like Tcerg1l 5730476P14Rik -165.9kb Anti-sense Chr. 7 - 138208972 - 138397730 on NCBI Build GRCm38
          Downstream mitogen-activated protein kinase 1 interacting protein 1 Mapk1ip1 2310009E07Rik,
          AU043776,
          MISS
          268.8kb Anti-sense Chr. 7 - 138835818 - 138846267 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 622377 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896648 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 507689 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 534730 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 93404 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959650 AK016032.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06389692 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06087015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06613150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06384910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06076879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06384903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06306211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06583382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06172589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06362494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06245521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06245519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06613145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06245523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25572711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06362502 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432274_PM_at - -
          Mouse Genome 430 2.0 Array 1432274_PM_at - -
          Mouse Genome 430 2.0 Array 1432274_at - -
          Mouse Expression Set 430 1432274_at - -
          Mouse Expression Set 430 1432274_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432274_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432274_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0