Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01277700_m1
          See other C230079O03RIK GE Assays ›
          Gene Symbol:
          C230079O03Rik
          Gene Name:
          RIKEN cDNA C230079O03 gene
          Gene Aliases:
          C230079O03
          Chromosome Location:
          Chr.7: 136332216 - 136349415 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01277700_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C230079O03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040459.1 2 - 3 121 144
          GenBank mRNA AK048899.1 - 2 - 3 122 144
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB651410.1 - 2 - 3 122 Chr. 7 - 136332993 - 136349415 [-] on Build GRCm38 144

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA C030029H02 gene C030029H02Rik -29.3kb Anti-sense Chr. 7 - 136268324 - 136318511 on NCBI Build GRCm38
          Downstream predicted gene 6249 Gm6249 EG621677 319.1kb Sense Chr. 7 - 136667955 - 136710604 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004456.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670102 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 217998 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 319395 AK135860.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 319395 BC013491.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 319395 NM_001033243.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969747 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 399110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 80741 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06384202 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112492 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06384202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06000398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06279856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06479413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06479407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05903821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05903819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05903822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06050728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25558592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06050718 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700037413.mm.1 TC0700004456.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037413.mm.1 NR_040459.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037413.mm.1 NR_040459.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037412.mm.1 TC0700004456.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037412.mm.1 NR_040459.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037412.mm.1 NR_040459.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037414.mm.1 TC0700004456.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037414.mm.1 NR_040459.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037414.mm.1 NR_040459.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 305400 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102810451 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0