Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01277832_m1
          See other SERPINE2 GE Assays ›
          Gene Symbol:
          Serpine2
          Gene Name:
          serine (or cysteine) peptidase inhibitor, clade E, member 2
          Gene Aliases:
          B230326M24Rik, PAI-1, PI-7, PI7, PN-1, Spi4
          Chromosome Location:
          Chr.1: 79794319 - 79861175 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01277832_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpine2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009255.4 NP_033281.1 5 - 6 1057 73
          GenBank mRNA AK045954.1 - 5 - 6 1029 73
          AK047856.1 - 5 - 6 1029 73
          AK051332.1 - 5 - 6 1029 73
          AK159687.1 - 5 - 6 1029 73
          BC010675.1 - 5 - 6 1007 3595407 73
          CT010311.1 - 4 - 5 880 73
          X70296.1 - 5 - 6 1028 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002645.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpine2

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          nervous system development
          blood coagulation
          negative regulation of cell population proliferation
          response to wounding
          negative regulation of peptidase activity
          negative regulation of platelet activation
          negative regulation of plasminogen activation
          negative regulation of sodium ion transport
          negative regulation of endopeptidase activity
          negative regulation of protein processing
          positive regulation of neuron projection development
          negative regulation of phosphatidylinositol 3-kinase signaling
          cerebellar granular layer morphogenesis
          cell differentiation
          negative regulation of blood coagulation
          negative regulation of cell growth
          secretion by cell
          secretory granule organization
          negative regulation of protein catabolic process
          mating plug formation
          negative regulation of proteolysis
          negative regulation of smoothened signaling pathway
          regulation of timing of cell differentiation
          positive regulation of astrocyte differentiation
          detection of mechanical stimulus involved in sensory perception
          regulation of synaptic transmission, glutamatergic
          long-term synaptic potentiation
          innervation
          seminal vesicle epithelium development
          dense core granule biogenesis
          negative regulation of platelet aggregation
          serine-type endopeptidase inhibitor activity
          signaling receptor binding
          glycosaminoglycan binding
          heparin binding
          peptidase inhibitor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100021527.mm.1 TC0100002645.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100021527.mm.1 NM_009255.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100021527.mm.1 NM_009255.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix