Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01277903_m1
          See other 1700003G18RIK GE Assays ›
          Gene Symbol:
          1700003G18Rik
          Gene Name:
          RIKEN cDNA 1700003G18 gene
          Gene Aliases:
          2610312K03Rik
          Chromosome Location:
          Chr.7: 116081762 - 116093149 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01277903_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700003G18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029433.1 3 - 4 296 145
          GenBank mRNA AK005637.1 - 309 145
          BC048672.1 - 3 - 4 289 145

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004151.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 893925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13829 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 829089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891665 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 361726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 645801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 114006 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145386 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06334189 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145386 - -
          Whole Mouse Genome G4122A A_51_P229230 BC048672.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P229230 NR_029433.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P229230 AK005637.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139593 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120292 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25446019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25446011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25446020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25446009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25446016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25446017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05959952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06127596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25446014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06543197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25446005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25446021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25446003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25446004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25446012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25446025 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431414_PM_at - -
          Mouse Genome 430 2.0 Array 1431414_at - -
          Mouse Genome 430 2.0 Array 1431414_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700034390.mm.1 TC0700004151.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700034390.mm.1 NR_029433.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034390.mm.1 NR_029433.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034391.mm.1 TC0700004151.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700034391.mm.1 NR_029433.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034391.mm.1 NR_029433.1 Refseq
          Mouse Expression Set 430 1431414_PM_at - -
          Mouse Expression Set 430 1431414_at - -
          Murine Genome U74v2 Set 168172_PM_r_at - -
          Murine Genome U74v2 Set 168172_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1431414_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431414_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 728173 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103780102 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0