Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01278905_g1
          See other DAXX GE Assays ›
          Gene Symbol:
          Daxx
          Gene Name:
          Fas death domain-associated protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 33909381 - 33915590 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01278905_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Daxx
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199733.1 NP_001186662.1 6 - 7 2212 78
          NM_007829.4 NP_031855.3 6 - 7 2112 78
          GenBank mRNA AF006040.1 - 5 - 6 1963 78
          AK090356.1 - 7 - 8 1923 78
          AK133863.1 - 6 - 7 2020 78
          AK144332.1 - 6 - 7 2045 78
          AK145906.1 - 6 - 7 2043 78
          AK146743.1 - 6 - 7 2039 78
          AK151279.1 - 6 - 7 2033 78
          AK153149.1 - 6 - 7 2000 78
          AK156425.1 - 6 - 7 2210 78
          AK172616.1 - 6 - 7 2213 78
          AY734459.1 - 4 - 5 1676 78
          AY734464.1 - 1 - 2 285 78
          AY734465.1 - 4 - 5 1587 78
          AY734466.1 - 1867 78
          AY758274.1 - 1 - 2 418 78
          BC128373.1 - 6 - 7 2022 40111681 78
          BQ176437.1 - 2 - 3 78
          KJ650207.1 - 5 - 6 1939 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000600.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Daxx

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          mitotic cytokinesis
          positive regulation of protein phosphorylation
          chromatin organization
          nucleosome assembly
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          apoptotic process
          negative regulation of myotube differentiation
          androgen receptor signaling pathway
          PML body organization
          regulation of protein ubiquitination
          positive regulation of histone phosphorylation
          positive regulation of apoptotic process
          positive regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          apoptotic signaling pathway
          positive regulation of neuron death
          negative regulation of nucleic acid-templated transcription
          positive regulation of apoptotic signaling pathway
          p53 binding
          transcription coactivator activity
          transcription corepressor activity
          protein binding
          transcription factor binding
          enzyme binding
          protein kinase binding
          protein kinase activator activity
          ubiquitin protein ligase binding
          histone binding
          protein homodimerization activity
          protein N-terminus binding
          androgen receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700005526.mm.1 TC1700000600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005526.mm.1 NM_001199733.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005526.mm.1 NM_001199733.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005526.mm.1 NM_007829.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005526.mm.1 NM_007829.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005528.mm.1 TC1700000600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005528.mm.1 NM_001199733.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005528.mm.1 NM_001199733.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005528.mm.1 NM_007829.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005528.mm.1 NM_007829.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0