Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01279053_m1
          See other SYCE1 GE Assays ›
          Gene Symbol:
          Syce1
          Gene Name:
          synaptonemal complex central element protein 1
          Gene Aliases:
          4933406J07Rik
          Chromosome Location:
          Chr.7: 140777229 - 140787854 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01279053_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Syce1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001143765.1 NP_001137237.1 2 - 3 187 154
          GenBank mRNA AK016694.1 - 2 - 3 187 154
          BY081845.1 - 2 - 3 188 154

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004504.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Recommended Assay Mus musculus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle
          meiotic prophase I
          synaptonemal complex assembly
          cell division
          meiotic cell cycle
          synaptonemal complex organization
          protein binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P444137 NM_001143765.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P444137 AK016694.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P444137 NM_001143765.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P444137 AK016694.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 506703 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741874 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 324052 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264407 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 859771 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402160 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 40007 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143218 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128849 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P444137 NM_001143765.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P444137 AK016694.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P444137 NM_001143765.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P444137 AK016694.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128849 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143218 - -
          Whole Mouse Genome G4122A A_51_P444137 NM_001143765.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P444137 AK016694.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P444137 NM_001143765.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P444137 AK016694.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143218 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05963880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06258267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25584913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25584906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25584932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25584920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06258272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25584916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25584909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06612706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25584903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25584923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06612707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25584926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25584933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06044505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25584929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25584919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06258260 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568977 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568981 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568979 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568971 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568980 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568976 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568978 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568973 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568974 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568975 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568970 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568969 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568972 - -
          Affymetrix HT MG-430 PM Array Plate 1428685_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10568968 - -
          Mouse Genome 430 2.0 Array 1428685_PM_at - -
          Mouse Genome 430 2.0 Array 1428685_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74075_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74075_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700037830.mm.1 TC0700004504.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037830.mm.1 NM_001143765.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037830.mm.1 NM_001143765.1 Refseq
          Mouse Expression Set 430 1428685_PM_at - -
          Mouse Expression Set 430 1428685_at - -
          Murine Genome U74v2 Set 168750_PM_i_at - -
          Murine Genome U74v2 Set 165885_at - -
          Murine Genome U74v2 Set 165885_PM_at - -
          Murine Genome U74v2 Set 168750_i_at - -
          Affymetrix HT Mouse Genome 430B Array 1428685_at - -
          Affymetrix HT Mouse Genome 430B Array 1428685_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7f58f9545f-46b6w:80/100.66.130.62:80.
          git-commit: c36c52929dfaa9f2956040e6abd275853d6e1b95
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline