Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01280429_s1
          See other PTCHD4 GE Assays ›
          Gene Symbol:
          Ptchd4
          Gene Name:
          patched domain containing 4
          Gene Aliases:
          3110082D06Rik, Gm321
          Chromosome Location:
          Chr.17: 42315810 - 42507742 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01280429_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptchd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028474.1 NP_082750.1 4 3393 156
          GenBank mRNA AK034360.1 - 1 470 156

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000822.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          hedgehog receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161257 NM_028474.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161257 AK034360.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161257 NM_028474.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161257 AK034360.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040227 BC151166.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040227 NM_028474.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2149670 AK014270.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171387 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040227 BC151166.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040227 NM_028474.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161257 NM_028474.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161257 AK034360.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161257 NM_028474.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161257 AK034360.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P415207 AK014270.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161257 NM_028474.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161257 AK034360.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161257 NM_028474.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161257 AK034360.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31184575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13263244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13111055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31184549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13546626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31184574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13263250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31184551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13111050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31184553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13546631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31184564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31184547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31184572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13274932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13263254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31184565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31184538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31184560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31184568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31184556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31184535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31184562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13111044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31184545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31183530 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445233 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445238 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445234 - -
          Affymetrix HT MG-430 PM Array Plate 1441559_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1453598_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10445232 - -
          Affymetrix Mouse Gene 1.1 ST Array 10445237 - -
          Mouse Genome 430 2.0 Array 1441559_at - -
          Mouse Genome 430 2.0 Array 1441559_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 627626_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 627626_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700007949.mm.1 TC1700000822.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007949.mm.1 NM_028474.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007949.mm.1 NM_028474.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007948.mm.1 TC1700000822.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007948.mm.1 NM_028474.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007948.mm.1 NM_028474.1 Refseq
          Mouse Expression Set 430 1441559_PM_at - -
          Mouse Expression Set 430 1441559_at - -
          Affymetrix HT Mouse Genome 430B Array 1441559_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441559_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 568030 - -
          ABI Mouse Genome Survey Microarray 849991 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103840176 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213239 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix