Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01280765_m1
          See other 4930441J16RIK GE Assays ›
          Gene Symbol:
          4930441J16Rik
          Gene Name:
          RIKEN cDNA 4930441J16 gene
          Gene Aliases:
          Astx1a, Astx1b, Astx1c
          Chromosome Location:
          Chr.2: 74298940 - 74307717 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01280765_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930441J16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040501.1 4 - 5 371 147
          GenBank mRNA AK015352.1 - 4 - 5 371 147
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY714907.1 - 4 - 5 368 Chr. 2 - 74298938 - 74307714 [-] on Build GRCm38 147

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ATP synthase,
          H+ transporting,
          mitochondrial F0 complex,
          subunit C3 (subunit 9)
          Atp5g3 6030447M23 -387.6kb Sense Chr. 2 - 73908447 - 73911326 on NCBI Build GRCm38
          Downstream limb and neural patterns Lnp 2310011O18Rik,
          4921514L11Rik,
          9530051D01Rik,
          AI666268,
          Ul,
          ulnaless
          214.3kb Sense Chr. 2 - 74514836 - 74579466 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003771.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01688641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01688635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01783852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01679542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01987739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01688644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01189021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21336705 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200032267.mm.1 TC0200003771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200032267.mm.1 NR_040501.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200032267.mm.1 NR_040501.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200032268.mm.1 TC0200003771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200032268.mm.1 NR_040501.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200032268.mm.1 NR_040501.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 368991 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0